Skip to Content
Merck
All Photos(3)

Key Documents

323543

Sigma-Aldrich

Europium(III) oxide

99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Eu2O3
CAS Number:
Molecular Weight:
351.93
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99.999% trace metals basis

form

powder and chunks

reaction suitability

reagent type: catalyst
core: europium

density

7.42 g/mL at 25 °C (lit.)

SMILES string

O=[Eu]O[Eu]=O

InChI

1S/2Eu.3O

InChI key

RSEIMSPAXMNYFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Priyam Singh et al.
Nanoscale, 9(2), 696-705 (2016-12-14)
We herein demonstrate novel inorganic-organic hybrid nanoparticles (HNPs) composed of inorganic NPs, NaY

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service