Skip to Content
Merck
All Photos(4)

Key Documents

310697

Sigma-Aldrich

Cellulose

microcrystalline, powder, 20 μm

Synonym(s):

Cellulose powder, Cotton linters

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

form

microcrystalline
powder

particle size

20 μm

pH

5-7 (11 wt. %)

bulk density

0.5 g/mL (25°C)

SMILES string

O1[C@H](C(C(C(C1CO)O)O)O)O[C@@H]2C(OC(C(C2O)O)O)CO

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3?,4?,5?,6?,7?,8?,9?,10-,11?,12+/m1/s1

InChI key

GUBGYTABKSRVRQ-WFVLMXAXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bulking agent, opacifier, anti-caking agent, extrusion aid and stabilizer for foams and emulsions.
High purity cellulose powders for partition chromatography.

Features and Benefits

Amorphous regions are hydrolyzed leaving crystalline microfibrils. Forms thixotropic gels, good thermal stability.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reza Amin et al.
Micromachines, 11(6) (2020-07-01)
To transform from reactive to proactive healthcare, there is an increasing need for low-cost and portable assays to continuously perform health measurements. The paper-based analytical devices could be a potential fit for this need. To miniaturize the multiplex paper-based microfluidic
Marco Mattonai et al.
Bioresource technology, 270, 270-277 (2018-09-18)
A combined study of crystallinity index (CI), degree of polymerisation (DP) and thermal stability of cellulose was carried out for monitoring the effect of ball-milling. DP and CI are two fundamental quantities that describe the physico-chemical behaviour of cellulose. Milling
Inka Vanwonterghem et al.
The ISME journal, 8(10), 2015-2028 (2014-04-18)
A replicate long-term experiment was conducted using anaerobic digestion (AD) as a model process to determine the relative role of niche and neutral theory on microbial community assembly, and to link community dynamics to system performance. AD is performed by
Eshchar Mizrachi et al.
The New phytologist, 194(1), 54-62 (2012-04-05)
Fast-growing, short-rotation forest trees, such as Populus and Eucalyptus, produce large amounts of cellulose-rich biomass that could be utilized for bioenergy and biopolymer production. Major obstacles need to be overcome before the deployment of these genera as energy crops, including
Rachel A Burton et al.
Current opinion in biotechnology, 26, 79-84 (2014-04-01)
Plant cell walls consist largely of cellulose, non-cellulosic polysaccharides and lignin. Concerted attempts are underway to convert wall polysaccharides from crop plant residues into renewable transport fuels and other valuable products, and to exploit the dietary benefits of cereal grain

Questions

1–4 of 4 Questions  
  1. When I checked the SDS, the cellulose content was less than 100%. What is the exact purity of the cellulose?

    1 answer
    1. The purity of this product has not been determined. However, as this material is prepared from wood, residual elements remaining from the manufacturing process may be present. In addition, the maximum allowable moisture content is 5.0% and is reported on the lot specific Certificate of Analysis.

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  4. Does the particle size refer to the length or diameter? Are these particles predominantly spherical or cylindrical?

    1 answer
    1. The particle size and morphology are not determined for this product. Semi-spherical particles may be assumed. Historical information indicates particles are <50 μm with an average size of 20 μm. The morphology may be confirmed by the end user via microscopy.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service