Skip to Content
Merck
All Photos(1)

Key Documents

309494

Sigma-Aldrich

Dibutyl adipate

96%

Synonym(s):

Adipic acid dibutyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH2CO2(CH2)3CH3]2
CAS Number:
Molecular Weight:
258.35
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

96%

form

liquid

refractive index

n20/D 1.436 (lit.)

bp

305 °C (lit.)

density

0.962 g/mL at 25 °C (lit.)

SMILES string

CCCCOC(=O)CCCCC(=O)OCCCC

InChI

1S/C14H26O4/c1-3-5-11-17-13(15)9-7-8-10-14(16)18-12-6-4-2/h3-12H2,1-2H3

InChI key

XTJFFFGAUHQWII-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibutyl adipate (DBA) is a plasticizer and a skin conditioning agent in cosmetics that is a mixture of a diester of butyl alcohol and adipic acid. It is soluble in water and organic solvents and does not absorb radiation in the ultraviolet region.

Application

DBA can be used as a plasticizer to produce a mixture with polyvinyl chloride (PVC), which forms soft gel type electro-active materials for soft robotic actuators and adaptive lenses. It may also be used to plasticize polyurethane gels for potential usage in biomedical applications.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adaptive Lenses Based on Soft Electroactive Materials.
Ren H and Wu S
Applied Sciences, 8(7), 1085-1085 (2018)
Cosmetics
Rieger MM
Kirk-Othmer Encyclopedia of Chemical Technology, 1-53 (2000)
Electrically Triggered Actuation of Plasticized Thermoplastic Polyurethane Gels.
Xia H, et al.
Macromolecular Materials and Engineering, 301(7), 864-869 (2016)
Electrical properties and electromechanical modeling of plasticized PVC gel actuators.
Asaka K and Hashimoto M
Sensors and Actuators B, Chemical, 273, 1246-1256 (2018)
Towards electroactive gel artificial muscle structures.
Helps T, et al.
Proceedings of SPIE, 10594, 1059408-1059408 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service