30740
Dehydroacetic acid
≥98.0% (T)
Synonym(s):
2-Acetyl-5-hydroxy-3-oxo-4-hexenoic acid δ-lactone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H8O4
CAS Number:
Molecular Weight:
168.15
Beilstein:
6129
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥98.0% (T)
form
powder
bp
270 °C (lit.)
mp
111-113 °C (lit.)
functional group
ester
ketone
SMILES string
CC(=O)C1C(=O)OC(C)=CC1=O
InChI
1S/C8H8O4/c1-4-3-6(10)7(5(2)9)8(11)12-4/h3,7H,1-2H3
InChI key
PGRHXDWITVMQBC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Dehydroacetic acid is one of the food additives found in red wine and was determined by ultra-fast liquid chromatography-tandem quadrupole mass spectrometry (UFLC-MS/MS).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
314.6 °F
Flash Point(C)
157 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Ségolène De Vaugelade et al.
Rapid communications in mass spectrometry : RCM, 32(11), 862-870 (2018-03-10)
The present work is devoted to the structural elucidation of by-products issued from the direct ultraviolet-visible (UV-vis) irradiation of dehydroacetic acid (DHA) in solution and in cosmetic emulsion. Analyses were carried out using gas chromatography coupled with ion trap mass
Xiao-Hong Chen et al.
Journal of chromatography. A, 1263, 34-42 (2012-10-10)
A novel and effective dispersive solid-phase extraction (dSPE) procedure with rapid magnetic separation using ethylenediamine-functionalized magnetic polymer as an adsorbent was developed. The new procedure had excellent clean-up ability for the selective removal of the matrix in red wine. An
Luiz C Dias et al.
Journal of agricultural and food chemistry, 57(4), 1399-1405 (2009-01-28)
A short and efficient approach to a range of new chiral and achiral functionalized (E)-enaminopyran-2,4-diones starting with commercially available dehydroacetic acid is described. The phytotoxic properties of these (E)-enaminopyran-2,4-diones were evaluated by their ability to interfere with the growth of
C S Mantzoros et al.
American journal of hypertension, 8(6), 606-614 (1995-06-01)
To test the hypothesis that androgens may play a role in the pathogenesis of atherosclerosis and hypertension, we investigated the association of delta 5 dehydroepiandrosterone sulfate [DHEAS]) and delta 4 (free testosterone [FT]) androgens with the resting, seated blood pressure
Yuka Sakaguchi et al.
Journal of applied toxicology : JAT, 28(4), 524-529 (2007-10-18)
Sodium dehydroacetate (DHA-S) is used as a food additive, preservative and antimicrobial agent. Repeated oral administration of DHA-S in rats induced severe hemorrhage in multiple organs and prolongation of blood coagulation factors. To determine the mechanism of hemorrhage, the protective
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service