Skip to Content
Merck
All Photos(2)

Documents

303518

Sigma-Aldrich

Tantalum(V) oxide

99% trace metals basis

Synonym(s):

Tantalum pentoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ta2O5
CAS Number:
Molecular Weight:
441.89
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: tantalum

density

8.2 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=[Ta](=O)O[Ta](=O)=O

InChI

1S/5O.2Ta

InChI key

PBCFLUZVCVVTBY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tantalum pentoxide is dielectric in nature making it useful for making capacitors in the electronics industry.

Application

Tantalum oxide is intermixed with silicon in gate dielectric structures. It may be used to synthesize hexagonally packed mesoporous tantalum oxide molecular sieves.Ion-beam sputter deposition of tantalum oxide films was investigated for possible optical coating applications.It may be used in making optical glass for lenses and in electronic circuits.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intermixing at the tantalum oxide/silicon interface in gate dielectric structures.
Alers GB, et al.
Applied Physics Letters, 73(11), 1517-1519 (1998)
Synthesis and characterization of hexagonally packed mesoporous tantalum oxide molecular sieves.
Antonelli DM and Ying JY
Chemistry of Materials, 8(4), 874-881 (1996)
Effects of oxygen content on the optical properties of tantalum oxide films deposited by ion-beam sputtering.
H Demiryont et al.
Applied optics, 24(4), 490-490 (1985-02-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service