Skip to Content
Merck
All Photos(2)

Key Documents

302163

Sigma-Aldrich

(R)-(−)-1-Phenyl-1,2-ethanediol

99%

Synonym(s):

(−)-Styrene glycol, (R)-(−)-Phenylethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(C6H5)OH
CAS Number:
Molecular Weight:
138.16
Beilstein:
2355547
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]20/D −69°, c = 1 in chloroform

bp

272-274 °C (lit.)

mp

64-67 °C (lit.)

SMILES string

OC[C@H](O)c1ccccc1

InChI

1S/C8H10O2/c9-6-8(10)7-4-2-1-3-5-7/h1-5,8-10H,6H2/t8-/m0/s1

InChI key

PWMWNFMRSKOCEY-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Intermediate for the synthesis of chiral phosphine catalysts for asymmetric hydrogenations.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Chem. Soc. Perkin Trans. II, 489-489 (1982)
The Journal of Organic Chemistry, 44, 1729-1729 (1979)
Yawei Geng et al.
Wei sheng wu xue bao = Acta microbiologica Sinica, 50(1), 60-66 (2010-03-30)
A novel (S)-specific carbonyl reductase gene (scr II) was cloned from the genome of Candida parapsilosis CCTCC M203011, and its catalytic function for the biotransformation of chiral alcohol was verified. The possible carbonyl reductase gene scr II was amplified by
Yao Nie et al.
Organic & biomolecular chemistry, 9(11), 4070-4078 (2011-04-21)
The application of biocatalysis to the synthesis of chiral molecules is one of the greenest technologies for the replacement of chemical routes due to its environmentally benign reaction conditions and unparalleled chemo-, regio- and stereoselectivities. We have been interested in
Magdy Y Shamy et al.
Journal of environmental pathology, toxicology and oncology : official organ of the International Society for Environmental Toxicology and Cancer, 21(1), 57-61 (2002-04-06)
Styrene is a known mutagen and suspected carcinogen, used in the reinforced plastic industry. This study aims to identify the occurrence of DNA single strand breaks (SSBs) in workers exposed to styrene levels far below the recommended standards. We compared

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service