Skip to Content
Merck
All Photos(2)

Key Documents

301353

Sigma-Aldrich

2,6-Naphthalenedicarboxylic acid

95%

Synonym(s):

2,6-Naphthalic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(CO2H)2
CAS Number:
Molecular Weight:
216.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

>300 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)c1ccc2cc(ccc2c1)C(O)=O

InChI

1S/C12H8O4/c13-11(14)9-3-1-7-5-10(12(15)16)4-2-8(7)6-9/h1-6H,(H,13,14)(H,15,16)

InChI key

RXOHFPCZGPKIRD-UHFFFAOYSA-N

Gene Information

human ... PTPN1(5770)

Looking for similar products? Visit Product Comparison Guide

General description

Self-assembly of 2,6-naphthalenedicarboxylic acid into two-dimensionally ordered supramolecular structures was studied at the liquid-solid interface by scanning tunneling microscopy. Solvothermal reaction of Mn(II) and 2,6-naphthalenedicarboxylic acid in diethylformamide to form a 3D porous metal-organic framework generating 1D channels was studied.

Application

2,6-Naphthalenedicarboxylic acid was used in preparation of poly-2,6-naphthalenes via metal-catalyzed polymerization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong-Ying Gao et al.
Journal of the American Chemical Society, 136(27), 9658-9663 (2014-06-18)
Metal-catalyzed polymerization of 2,6-naphthalenedicarboxylic acid (NDCA) to form poly-2,6-naphthalenes at various surfaces is reported. Polymerizations occur via initial formal dehydrogenation of self-assembled diacids with subsequent decarboxylation to give polymeric bisnaphthyl-Cu species at elevated temperature as intermediate structures (<160 °C). Further
Hoi Ri Moon et al.
Inorganic chemistry, 45(21), 8672-8676 (2006-10-13)
A 3D porous metal-organic framework generating 1D channels, [Mn(NDC)(DEF)]n (1), has been prepared from the solvothermal reaction of Mn(II) and 2,6-naphthalenedicarboxylic acid (H2NDC) in diethylformamide (DEF). When DEF molecules coordinating Mn(II), which occupy the channels, are removed from 1 by
M A Strege et al.
Journal of chromatography. B, Biomedical sciences and applications, 697(1-2), 255-257 (1997-10-29)
A rapid and simple method for the capillary electrophoretic determination of residual trifluoroacetic acid in lyophilized natural products is described. The technique utilizes 2,6-naphthalenedicarboxylic acid as a separation buffer additive providing indirect ultraviolet absorption detection. Using this method, acceptable precision
Dong-Sung Kim et al.
Biotechnology letters, 30(2), 329-333 (2007-10-05)
Crude 2,6-naphthalene dicarboxylic acid was purified by Pseudomonas sp. HN-72 which biotransformed the major impurity of 2-formyl-6-naphthoic acid into 2,6-naphthalene dicarboxylic acid. The biotransformation yield reached 100% when the reaction was performed at 40 degrees C for 1 h, in
Christoph Heininger et al.
Langmuir : the ACS journal of surfaces and colloids, 25(2), 968-972 (2008-12-19)
Self-assembly into two-dimensionally ordered supramolecular structures of three aromatic dicarboxylic acids-2,6-naphthalenedicarboxylic acid (NDA), 4,4'-biphenyldicarboxylic acid (BPDA), and 4,4'-stilbenedicarboxylic acid (SDA)-is studied at the liquid-solid interface by scanning tunneling microscopy. All compounds possess structural similarities, namely, two interconnected aromatic moieties and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service