301248
Tris[2-(2-methoxyethoxy)ethyl]amine
95%
Synonym(s):
TDA-1, Tris(3,6-dioxaheptyl)amine
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
(CH3OCH2CH2OCH2CH2)3N
CAS Number:
Molecular Weight:
323.43
Beilstein:
2369296
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
95%
form
liquid
refractive index
n20/D 1.4486 (lit.)
density
1.011 g/mL at 25 °C (lit.)
functional group
amine
ether
SMILES string
COCCOCCN(CCOCCOC)CCOCCOC
InChI
1S/C15H33NO6/c1-17-10-13-20-7-4-16(5-8-21-14-11-18-2)6-9-22-15-12-19-3/h4-15H2,1-3H3
InChI key
XGLVDUUYFKXKPL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Tris[2-(2-methoxyethoxy)ethyl]amine was used:
- in oxidation of arylmethanols under phase-transfer conditions
- as cryptand during phase-transfer glycosylation of 2-amino-4-methoxy-7H-pyrrolo[2,3-d]pyrimidine
- as phase-transfer catalyst in synthesis of 1,3-dideaza-2′-deoxyadenosine and related benzimidazole 2′-deoxyribonucleosides
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1C
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
323.6 °F - closed cup
Flash Point(C)
162 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Liquid-liquid and solid-liquid phase-transfer glycosylation of pyrrolo [2, 3-d] pyrimidines: stereospecific synthesis of 2-deoxy-?-D-ribofuranosides related to 2'-deoxy-7-carbaguanosine.
Seela F, et al.
Journal of the Chemical Society. Perkin Transactions 1, 3, 697-702 (1998)
Phase-Transfer Catalysed Permanganate Oxidations Using Tris [2-(2-Methoxyethoxy) Ethyl] Amine (Tda-1).
Mckillop A and Mills LS.
Synthetic Communications, 17(6), 647-655 (1987)
Stereoselective glycosylation of nitrobenzimidazole anions: synthesis of 1, 3-dideaza-2'-deoxyadenosine and related 2'-deoxyribofuranosides.
Seela F and Bourgeois W.
Synthesis, 12, 912-918 (1989)
Falk Kunkel et al.
Molecules (Basel, Switzerland), 20(11), 20805-20822 (2015-11-28)
DNA methyltransferases (MTases) catalyze the transfer of the activated methyl group of the cofactor S-adenosyl-l-methionine (AdoMet or SAM) to the exocyclic amino groups of adenine or cytosine or the C5 ring atom of cytosine within specific DNA sequences. The DNA
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service