Skip to Content
Merck
All Photos(1)

Key Documents

292966

Sigma-Aldrich

1,1′-Binaphthyl-2,2′-diyl hydrogenphosphate

95%

Synonym(s):

BNDHP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H13O4P
CAS Number:
Molecular Weight:
348.29
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling

SMILES string

OP1(=O)Oc2ccc3ccccc3c2-c4c(O1)ccc5ccccc45

InChI

1S/C20H13O4P/c21-25(22)23-17-11-9-13-5-1-3-7-15(13)19(17)20-16-8-4-2-6-14(16)10-12-18(20)24-25/h1-12H,(H,21,22)

InChI key

JEHUZVBIUCAMRZ-UHFFFAOYSA-N

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Bielejewska et al.
Journal of chromatography. A, 977(2), 225-237 (2002-11-29)
The chiral recognition ability of single and dual selectors, that were used as additives, have been investigated by HPLC and CE. Native beta- and gamma-cyclodextrins, permethylated beta-cyclodextrin, hydroxypropyl-beta-cyclodextrin, cholic acid and taurodeoxycholic acid sodium salts were applied as chiral selectors
Yingying Dong et al.
Electrophoresis, 32(13), 1735-1741 (2011-06-28)
Gels formed by self-association of monomeric guanosine compounds join numerous other agents such as cyclodextrins, crown ethers, chiral surfactants, antibiotics, proteins, and polysaccharides for chiral separations. Guanosine gels (G-gels) are self-assembled networks of hydrogen-bonded tetrads formed by guanosine nucleotides and
Christine M Hebling et al.
Langmuir : the ACS journal of surfaces and colloids, 24(24), 13866-13874 (2008-12-05)
Bile salt micelles can be employed as a pseudostationary phase in micellar electrokinetic capillary chromatography (MEKC) separations of chiral analytes. To improve MEKC separations of chiral analytes, a molecular level understanding of micelle aggregation in the presence of analyte is
Ulrich Schmitt et al.
Electrophoresis, 25(16), 2801-2807 (2004-09-08)
Comparative enantioseparation of the enantiomers of 1,1'-binaphthyl-2,2'-diyl hydrogen phosphate was performed with cyclodextrin (CD)-modified capillary electrophoresis (CE). Two single isomers, beta-CD, heptakis(2,3,6-tri-O-methyl)-beta-CD (TM-beta-CD), and heptakis(2,6-di-O-methyl)-beta-CD (DM-beta-CD) of 98% purity as well as heptakis(2,3-di-O-acetyl)-beta-CD were used and compared in terms of
Stephanie A Kingsbury et al.
Magnetic resonance in chemistry : MRC, 48(3), 184-191 (2010-01-06)
NMR spectroscopy was used to characterize the binding of the chiral compound 1,1'-binaphthyl-2,2'-diyl hydrogen phosphate (BNP) to five molecular micelles with chiral dipeptide headgroups. Molecular micelles have covalent linkages between the surfactant monomers and are used as chiral mobile phase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service