Skip to Content
Merck
All Photos(1)

Key Documents

289388

Sigma-Aldrich

Silicon tetrachloride

99.998% trace metals basis

Synonym(s):

STC, Tetrachlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiCl4
CAS Number:
Molecular Weight:
169.90
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

vapor density

5.86 (vs air)

vapor pressure

420 mmHg ( 37.7 °C)

Assay

99.998% trace metals basis

form

liquid

reaction suitability

core: silicon

bp

57.6 °C (lit.)

mp

−70 °C (lit.)

density

1.483 g/mL at 25 °C (lit.)

SMILES string

Cl[Si](Cl)(Cl)Cl

InChI

1S/Cl4Si/c1-5(2,3)4

InChI key

FDNAPBUWERUEDA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Semiconductor grade

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Khushalani et al.
Journal of nanoscience and nanotechnology, 1(2), 129-132 (2003-08-14)
Mesostructured silica-nanoparticle monolithic composites have been synthesized by dispersing prefabricated nanoparticles of gold or zeolite (silicalite) in ethanolic reaction mixtures containing SiCl4 and a Pluronic triblock copolymer template. Whereas silicalite nanoparticles were used directly, surface functionalization of the gold nanoparticles
Pulmonary interstitial emphysema: CT findings.
A C Kemper et al.
AJR. American journal of roentgenology, 172(6), 1642-1642 (1999-06-01)
Claudio Curti et al.
Organic letters, 13(17), 4738-4741 (2011-08-13)
Virtually perfect transmittal of the enolate reactivity up to five conjugated double bonds from the origin allows a series of furan-based silyloxypolyenes to add to aldehyde carbonyls at the most distant point of the molecule. Denmark's axially chiral bisphosphoramide/SiCl(4) combination
Sang Ho Lee et al.
Journal of the American Chemical Society, 127(25), 9071-9078 (2005-06-23)
A series of four asymmetrically aryl-substituted 9,9'-spiro-9-silabifluorene (SSF) derivatives, 2,2'-di-tert-butyl-7,7'-diphenyl-9,9'-spiro-9-silabifluorene (PhSSF), 2,2'-di-tert-butyl-7,7'-dipyridin-2-yl-9,9'-spiro-9-silabifluorene (PySSF), 2,2'-di-tert-butyl-7,7'-dibiphenyl-4-yl-9,9'-spiro-9-silabifluorene (BPhSSF), and 2,2'-di-tert-butyl-7,7'-bis(2',2' '-bipyridin-6-yl)-9,9'-spiro-9-silabifluorene (BPySSF) are prepared through the cyclization of the corresponding 2,2'-dilithiobiphenyls with silicon tetrachloride. These novel spiro-linked silacyclopentadienes (siloles) form transparent and stable
T Kapias et al.
Journal of hazardous materials, 81(3), 209-222 (2001-02-13)
Silicon tetrachloride is a toxic, corrosive water reactive substance that is used widely in the process industries. On spillage from containment it creates liquid pools that can either boil or evaporate. The main feature of the pool behaviour is the

Articles

Flame spray synthesis allows the scalable fabrication of many mixed oxide, salt, metal, carbon-coated metal, silica-coated metal oxide or even metal-ceramic composite nanoparticles.

Flame spray synthesis allows the scalable fabrication of many mixed oxide, salt, metal, carbon-coated metal, silica-coated metal oxide or even metal-ceramic composite nanoparticles.

Flame spray synthesis allows the scalable fabrication of many mixed oxide, salt, metal, carbon-coated metal, silica-coated metal oxide or even metal-ceramic composite nanoparticles.

Flame spray synthesis allows the scalable fabrication of many mixed oxide, salt, metal, carbon-coated metal, silica-coated metal oxide or even metal-ceramic composite nanoparticles.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service