Skip to Content
Merck
All Photos(1)

Documents

286958

Sigma-Aldrich

Phenylpyruvic acid

98%

Synonym(s):

2-Oxo-3-phenylpropanoic acid, 2-Oxo-3-phenylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2COCOOH
CAS Number:
Molecular Weight:
164.16
Beilstein:
2207312
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

150-154 °C (lit.)

storage temp.

−20°C

SMILES string

OC(=O)C(=O)Cc1ccccc1

InChI

1S/C9H8O3/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5H,6H2,(H,11,12)

InChI key

BTNMPGBKDVTSJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenylpyruvic acid reduces glucose-6-phosphate dehydrogenase activity without pre-incubation.

Application

Phenylpyruvic acid was used in the synthesis of 3-phenyllactic acid (PLA) by lactate dehydrogenase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuhuai Yu et al.
Biotechnology letters, 36(3), 627-631 (2013-11-20)
3-Phenyllactic acid (PLA) is an antimicrobial compound with broad and effective antimicrobial activity against both bacteria and fungi. Enzymatic production of PLA can be carried out from phenylpyruvic acid by lactate dehydrogenase (LDH); however, the enzymatic reaction is accompanied by
Faqing Tang et al.
Clinical biochemistry, 44(8-9), 711-718 (2011-03-16)
To search for markers of nasopharyngeal carcinoma (NPC) for diagnosis. Using gas chromatography and mass spectrometry, we evaluated 51 serum metabolites in 49 NPC, 37 throat cancer patients and 40 healthy controls. High metabolites were selected and confirmed in NPC
Hajer Ouertatani-Sakouhi et al.
Journal of biomolecular screening, 15(4), 347-358 (2010-03-17)
Macrophage migration inhibitory factor (MIF) is a major mediator of innate immunity and inflammation and presents a potential therapeutic target for various inflammatory, infectious, and autoimmune diseases, including cancer. Although a number of inhibitors have been identified and designed based
Tapan Kanti Paine et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(21), 6073-6081 (2007-05-01)
Iron(II)-phenylpyruvate complexes of tetradentate tris(6-methyl-2-pyridylmethyl)amine (6-Me3-TPA) and tridentate benzyl bis(2-quinolinylmethyl)amine (Bn-BQA) were prepared to gain insight into C-C bond cleavage catalyzed by dioxygenase enzymes. The complexes we have prepared and characterized are [Fe(6-Me3-tpa)(prv)][BPh4] (1), [Fe2(6-Me3-tpa)2(pp)][(BPh4)2] (2), and [Fe2(6-Me3-tpa)2(2'-NO2-pp)][(BPh4)2] (3), [Fe(6-Me3-tpa)(pp-Me)][BPh4]
Satoshi Hirano et al.
The Journal of biological chemistry, 283(10), 6459-6466 (2008-01-04)
Violacein and the indolocarbazoles are naturally occurring bisindole products with various biological activities, including antitumor activity. Although these compounds have markedly different molecular skeletons, their biosynthetic pathways share the same intermediate "compound X," which is produced from L-tryptophan via indole-3-pyruvic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service