Skip to Content
Merck
All Photos(2)

Documents

280372

Sigma-Aldrich

1-Butylpyrrolidine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17N
CAS Number:
Molecular Weight:
127.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.44 (lit.)

bp

155-157 °C/754 mmHg (lit.)

density

0.814 g/mL at 25 °C (lit.)

SMILES string

CCCCN1CCCC1

InChI

1S/C8H17N/c1-2-3-6-9-7-4-5-8-9/h2-8H2,1H3

InChI key

JSHASCFKOSDFHY-UHFFFAOYSA-N

Application

1-Butylpyrrolidine has been used in:
  • microwave-assisted synthesis of ionic liquid precursor, 1-butyl-1-methylpyrrolidinium methylcarbonate
  • [N,N-methylbutylpyrrolidinium] thiosalicylate, ionic liquid

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

96.8 °F - closed cup

Flash Point(C)

36 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterization and thermal properties of thiosalicylate ionic liquids.
Wilfred CD and Mustafa FB.
Journal of Chemical Sciences (Bangalore), 125(6), 1511-1515 (2013)
Giridhar Pulletikurthi et al.
Dalton transactions (Cambridge, England : 2003), 46(2), 455-464 (2016-12-14)
The mixtures of 1-butylpyrrolidine and ZnCl
Giridhar Pulletikurthi et al.
Chemistry, an Asian journal, 12(20), 2684-2693 (2017-08-05)
Electrostatic interactions are characteristic of ionic liquids (ILs) and play a pivotal role in determining the formation of species when solutes are dissolved in them. The formation of new species/complexes has been investigated for certain ILs. However, such investigations have
Optimised microwave-assisted synthesis of methylcarbonate salts: a convenient methodology to prepare intermediates for ionic liquid libraries.
Holbrey JD, et al.
Green Chemistry, 12(3), 407-413 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service