Skip to Content
Merck
All Photos(1)

Key Documents

270512

Supelco

Heptane

HPLC grade, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH3
CAS Number:
Molecular Weight:
100.20
Beilstein:
1730763
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
grade:
HPLC grade
Assay:
≥99%
technique(s):
HPLC: suitable
bp:
98 °C (lit.)
98 °C
vapor pressure:
40 mmHg ( 20 °C)
83 mmHg ( 37.7 °C)

grade

HPLC grade

Quality Level

vapor density

3.5 (vs air)

vapor pressure

40 mmHg ( 20 °C)
83 mmHg ( 37.7 °C)

Assay

≥99%

form

liquid

autoignition temp.

433 °F

purified by

glass distillation

expl. lim.

7 %

technique(s)

HPLC: suitable

impurities

<0.010% water

evapn. residue

<0.0003%

refractive index

n20/D 1.387 (lit.)

bp

98 °C (lit.)
98 °C

mp

−91 °C (lit.)

solubility

water: insoluble

density

0.684 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 197 nm Amax: 1.00
λ: 210 nm Amax: 0.40
λ: 220 nm Amax: 0.15
λ: 230 nm Amax: 0.05
λ: 260-400 nm Amax: 0.01

SMILES string

CCCCCCC

InChI

1S/C7H16/c1-3-5-7-6-4-2/h3-7H2,1-2H3

InChI key

IMNFDUFMRHMDMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bacterial phospholipid molecular species analysis by ion-pair reversed-phase HPLC/ESI/MS.
Mazzella N, et al.
Journal of Lipid Research, 45(7), 1355-1363 (2004)
A comprehensive modeling study of n-heptane oxidation.
Curran HJ, et al.
Combustion and Flame, 114(1), 149-177 (1998)
Chris G C Jacobs et al.
Insect biochemistry and molecular biology, 60, 7-12 (2015-03-10)
Insects have been extraordinary successful in colonizing terrestrial habitats and this success is partly due to a protective cuticle that mainly contains chitin and proteins. The cuticle has been well studied in larvae and adults, but little attention has been
Arun K Kota et al.
Advanced materials (Deerfield Beach, Fla.), 24(43), 5838-5843 (2012-08-30)
Hierarchically structured, superoleophobic surfaces are demonstrated that display one of the lowest contact angle hysteresis values ever reported - even with extremely low-surface-tension liquids such as n-heptane. Consequently, these surfaces allow, for the first time, even ≈2 μL n-heptane droplets
Jovana Zečević et al.
Nature, 528(7581), 245-248 (2015-12-15)
The ability to control nanoscale features precisely is increasingly being exploited to develop and improve monofunctional catalysts. Striking effects might also be expected in the case of bifunctional catalysts, which are important in the hydrocracking of fossil and renewable hydrocarbon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service