Skip to Content
Merck
All Photos(2)

Documents

268569

Sigma-Aldrich

Trimethylaluminum solution

2.0 M in hexanes

Synonym(s):

Trimethylalane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3Al
CAS Number:
Molecular Weight:
72.09
Beilstein:
3587197
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

2.0 M in hexanes

density

0.683 g/mL at 25 °C

SMILES string

C[Al](C)C

InChI

1S/3CH3.Al/h3*1H3;

InChI key

JLTRXTDYQLMHGR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trimethylaluminum is a Lewis acid having wide applications in organic synthesis. It is employed in organic reactions like cross-coupling reaction, methylenation, carboalumination, cyclopropanation, and rearrangement reactions. Tebbe Reagent (methylenation agent) can be prepared in situ by reacting trimethylaluminum with dichlorobis(cyclopentadienyl)titanium.

Application

Used with 4-bromo-2,6-di-tert-butylphenol (cat. no. 394378) to prepare a wide range of carbonyl compounds via the selective rearrangement of epoxides.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Repr. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trimethylaluminum
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2007)
Tetrahedron Letters, 30, 5607-5607 (1989)
Tetrahedron, 47, 6983-6983 (1991)
Mahtab Farzin et al.
Pathology, 47(4), 302-307 (2015-05-06)
BRCA1-associated protein 1 (BAP1) is a tumour suppressor gene frequently inactivated in mesothelioma, rarely also in association with germline mutation. BAP1 mutations have been associated with improved prognosis and distinct clinicopathological features. We sought to determine the clinicopathological significance of
Patricia A Mote et al.
Breast cancer research and treatment, 151(2), 309-318 (2015-04-29)
Progesterone receptor (PR) function, while essential in normal human breast, is also implicated in breast cancer risk. The two progesterone receptors, PRA and PRB, are co-expressed at equivalent levels in normal breast, but early in carcinogenesis normal levels of PRA:PRB

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service