Skip to Content
Merck
All Photos(2)

Key Documents

265470

Sigma-Aldrich

Bismuth

shot, 4-30 mesh, 99.9% trace metals basis

Synonym(s):

Bismuth element, Bismuth-209

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Bi
CAS Number:
Molecular Weight:
208.98
EC Number:
MDL number:
UNSPSC Code:
12141706
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

99.9% trace metals basis

form

shot

resistivity

129 μΩ-cm, 20°C

particle size

4-30 mesh

bp

1560 °C (lit.)

mp

271 °C (lit.)

density

9.8 g/mL at 25 °C (lit.)

SMILES string

[Bi]

InChI

1S/Bi

InChI key

JCXGWMGPZLAOME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Philip C Andrews et al.
Chemical communications (Cambridge, England), 49(28), 2870-2872 (2013-03-02)
Reaction of Bi(O(t)Bu)3 with aspirin (acetylsalicylic acid = aspH) in dry toluene results in the bismuth(III) complex, [Bi(O2C(C6H4)OAc)3]∞ 1 (O2C(C6H4)OAc = asp), minimum inhibitory concentration (MIC) against Helicobacter pylori ≥ 6.25 μg mL(-1), while the inclusion of a stoichiometric equivalent
Julie Pannequin et al.
The Journal of biological chemistry, 279(4), 2453-2460 (2003-10-08)
Although bismuth salts have been used for over two centuries for the treatment of various gastrointestinal disorders, the mechanism of their therapeutic action remains controversial. Because gastrins bind two trivalent ferric ions with high affinity, and because ferric ions are
A Slikkerveer et al.
Medical toxicology and adverse drug experience, 4(5), 303-323 (1989-09-01)
Inorganic bismuth salts are poorly soluble in water: solubility is influenced by the acidity of the medium and the presence of certain compounds with (hydr)oxy or sulfhydryl groups. The analysis of bismuth in biological material is not standardised and is
Jia-Tao Sun et al.
Physical review letters, 109(24), 246804-246804 (2013-02-02)
Scanning tunnelling microscopy and spectroscopy experiments complemented by first-principles calculations have been conducted to study the electronic structure of 4 monolayer Bi(110) nanoribbons on epitaxial graphene on silicon carbide [4H-SiC(0001)]. In contrast with the semimetal property of elemental bismuth, an
B Arnaud et al.
Physical review letters, 110(1), 016405-016405 (2013-02-07)
By means of first principles calculations, we compute the effective electron-phonon coupling constant G(0) governing the electron cooling in photoexcited bismuth. G(0) strongly increases as a function of electron temperature, which can be traced back to the semimetallic nature of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service