Skip to Content
Merck
All Photos(1)

Key Documents

264970

Sigma-Aldrich

Didecylamine

95%

Synonym(s):

N,N-Didecylamine, N-Decyl-1-decanamine, N-Decyldecanamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)9]2NH
CAS Number:
Molecular Weight:
297.56
Beilstein:
1763492
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

bp

179-180 °C/2 mmHg (lit.)

mp

38-40 °C (lit.)

SMILES string

CCCCCCCCCCNCCCCCCCCCC

InChI

1S/C20H43N/c1-3-5-7-9-11-13-15-17-19-21-20-18-16-14-12-10-8-6-4-2/h21H,3-20H2,1-2H3

InChI key

GMTCPFCMAHMEMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David P Long et al.
Journal of the American Chemical Society, 129(49), 15303-15310 (2007-11-14)
We compare the low-temperature electron transport properties of alkyl monolayers which utilize different attachment strategies to gold. Inelastic electron tunneling spectroscopy (IETS) and current-voltage analysis were performed on molecular junctions incorporating alkyl-dithiocarbamate and alkanethiolate self-assembled monolayers of similar length. Alkyl-dithiocarbamate
Dithiocarbamate-protected ruthenium nanoparticles: synthesis, spectroscopy, electrochemistry and STM studies.
Chen W, et al.
Electrochimica Acta, 53(3), 1150-1156 (2007)
Investigation into extraction of metal ion complexes with 1-phenyl-3-amino-5-pyrazolone-4-dithiocarboxylic acid.
Rudzit GP and Baltagalve IV.
Izv. Akad. Nauk, Ser. Khim, 6, 719-723 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service