All Photos(1)
About This Item
Linear Formula:
BrC6H3F2
CAS Number:
Molecular Weight:
192.99
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
refractive index
n20/D 1.505 (lit.)
bp
150-151 °C (lit.)
density
1.707 g/mL at 25 °C (lit.)
functional group
bromo
fluoro
SMILES string
Fc1ccc(Br)cc1F
InChI
1S/C6H3BrF2/c7-4-1-2-5(8)6(9)3-4/h1-3H
InChI key
YMQPKONILWWJQG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Regioselective nucleophilic aromatic substitution reaction of 4-bromo-1,2-difluorobenzene with benzyl alcohol has been investigated.
Application
4-Bromo-1,2-difluorobenzene has been used in the preparation of 4-benzyl-2-(3,4-difluorophenyl)-2-hydroxy-morpholine.
accessory
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
91.4 °F
Flash Point(C)
33 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Regioselectivity of fluorine substitution by alkoxides on unsymmetrical difluoroarenes.
Dirr R, et al.
Tetrahedron Letters, 49(31), 4588-4590 (2008)
Preparation of chiral key intermediates of morpholine based neurokinin receptor antagonists by asymmetric allylic alkylation.
Keldenich J, et al.
Tetrahedron, 69(31), 6424-6430 (2013)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service