Skip to Content
Merck
All Photos(1)

Documents

258083

Sigma-Aldrich

4-Bromobutyric acid

98%

Synonym(s):

4-Bromobutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)3COOH
CAS Number:
Molecular Weight:
167.00
Beilstein:
1743056
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

reaction suitability

reagent type: cross-linking reagent

bp

128-131 °C/11 mmHg (lit.)

mp

30-31 °C (lit.)

functional group

bromo
carboxylic acid

SMILES string

BrCCCC(O)=O

InChI

1S/C4H7BrO2/c5-3-1-2-4(6)7/h1-3H2,(H,6,7)

InChI key

GRHQDJDRGZFIPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Montanari et al.
Carbohydrate polymers, 221, 209-220 (2019-06-23)
Hyaluronan (HA) is among the most used biopolymers for viscosupplementation and dermocosmetics. However, the current injectable HA-based formulations present relevant limitations: I) unmodified HA is quickly degraded by endogenous hyaluronidases (HAase), resulting in short lasting properties; II) cross-linked HA, although
E Montanari et al.
Journal of controlled release : official journal of the Controlled Release Society, 326, 1-12 (2020-06-20)
Intracellular pathogens are a critical challenge for antimicrobial therapies. Staphylococcus aureus (S. aureus) causes approximately 85% of all skin and soft tissue infections in humans worldwide and more than 30% of patients develop chronic or recurrent infections within three months
Elita Montanari et al.
Advanced healthcare materials, 7(12), e1701483-e1701483 (2018-04-27)
Staphylococcus aureus is one of the most significant human pathogens that is frequently isolated in a wide range of superficial and systemic infections. The ability of S. aureus to invade and survive within host cells such as keratinocytes and host
Amelia C McCue et al.
Photochemistry and photobiology, 94(3), 545-551 (2018-01-30)
Light-responsive compounds have been used to manipulate biological systems with spatial and temporal control of the event of interest. Illumination of alkylcobalamins with green light (>500 nm) produces carbon-centered radicals, which have been demonstrated to effectively cause DNA damage. Molecules that
Renwu Li et al.
Bioorganic & medicinal chemistry, 28(7), 115356-115356 (2020-02-19)
Past few years have seen an active pursuit of the inhibitors for the deacylation catalyzed by the seven human sirtuins (i.e. SIRT1-7) as valuable chemical biological/pharmacological probes of this enzymatic deacylation and lead compounds for developing novel therapeutics for human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service