Skip to Content
Merck
All Photos(1)

Key Documents

254916

Sigma-Aldrich

7,8-Dihydroxy-6-methoxycoumarin

98%

Synonym(s):

Fraxetin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O5
CAS Number:
Molecular Weight:
208.17
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

powder

mp

230-231 °C (lit.)

SMILES string

COc1cc2C=CC(=O)Oc2c(O)c1O

InChI

1S/C10H8O5/c1-14-6-4-5-2-3-7(11)15-10(5)9(13)8(6)12/h2-4,12-13H,1H3

InChI key

HAVWRBANWNTOJX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

7,8-Dihydroxy-6-methoxycoumarin is a versatile compound known for its unique fluorescence properties and its role as a photoinitiator in polymerization processes. It exhibits high reactivity under UV light, making it an ideal candidate for applications in the polymerization industry, particularly in the formulation of coatings, adhesives, and inks. It is also used in biomedical applications, such as drug delivery due to its biocompatibility.

Application

7,8-Dihydroxy-6-methoxycoumarincan be used as a UV absorber in polymer formulations. Its ability to absorbultraviolet light helps in protecting polymers from degradation caused by UVradiation, thereby improving their longevity and stability.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Po-Lin Kuo et al.
International immunopharmacology, 6(7), 1167-1175 (2006-05-23)
The survival of osteoblast cells is one of the determinants of the development of osteoporosis in patients with inflamed synovium, such as in rheumatoid arthritis (RA). By means of alkaline phosphatase (ALP) activity and osteocalcin ELISA assay, we have shown
Joana Terés et al.
Plant, cell & environment, 42(8), 2384-2398 (2019-04-25)
High soil carbonate limits crop performance especially in semiarid or arid climates. To understand how plants adapt to such soils, we explored natural variation in tolerance to soil carbonate in small local populations (demes) of Arabidopsis thaliana growing on soils
María Isabel Sánchez-Reus et al.
Neuroscience research, 53(1), 48-56 (2005-07-06)
Fraxetin belongs to an extensive group of natural phenolic anti-oxidants. In the present study, using a human neuroblastoma SH-SY5Y cells, we have investigated the protective effects of this compound on modifications in endogenous reduced glutathione (GSH), intracellular oxygen species (ROS)
Po-Lin Kuo et al.
Biological & pharmaceutical bulletin, 29(1), 119-124 (2006-01-06)
Fraxetin (7,8-dihydroxy-6-methoxy coumarin), a coumarin derivative, was investigated for its effects on differentiation of osteoblasts. By means of alkaline phosphatase (ALP) activity and osteocalcin ELISA assay, we have shown that fraxetin exhibits a significant induction of differentiation in two human
María Francisca Molina-Jiménez et al.
Brain research, 1009(1-2), 9-16 (2004-05-04)
Rotenone-induced apoptosis is considered to contribute to the etiology of Parkinson's disease (PD). We try to prevent the apoptosis induced by rotenone toxicity with 50 microM myricetin, 100 microM fraxetin and 100 microM N-acetylcysteine (NAC) that protect against reactive oxygen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service