Skip to Content
Merck
All Photos(2)

Documents

252832

Sigma-Aldrich

Tropone

97%

Synonym(s):

2,4,6-Cycloheptatrien-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6O
CAS Number:
Molecular Weight:
106.12
Beilstein:
1902335
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.615 (lit.)

bp

113 °C/15 mmHg (lit.)

density

1.094 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

O=C1C=CC=CC=C1

InChI

1S/C7H6O/c8-7-5-3-1-2-4-6-7/h1-6H

InChI key

QVWDCTQRORVHHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Metal-catalyzed [6+3] cycloaddition of tropone with azomethine ylides has been reported.

Application

Tropone has been used in synthesis of:
  • bicyclic δ-lactones via heterocyclic carbine-catalyzed [8+3] annulation pathway
  • 6,7-benzobicyclo [3.2.2] nona-3,6,8-trien-2-one via thermal addition to bezyne

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Carmen Carreño et al.
Chemical communications (Cambridge, England), (8)(8), 1007-1009 (2005-02-19)
Synthesis of 4-aminotropones through a cyclization-ring expansion process occurs in a single step and with excellent yields from 4-amino-2,5-cyclohexadienones (p-quinamines) bearing a 4-sulfinyl or sulfonyl methyl group.
Reaction of tropone with benzyne. Formation and photoisomerization of 6, 7-benzobicyclo [3.2. 2] nona-3, 6, 8-trien-2-one.
Ciabattoni J, et al.
Journal of the American Chemical Society, 89(11), 2778-2779 (1967)
Young-Sun Do et al.
The Journal of organic chemistry, 74(2), 917-920 (2008-12-05)
A ring-expansion protocol that consisted of the 1,2-addition of various enolate nucleophiles to 6-trimethylsiloxy-2-cyclohexene-1-one (1) and the NaIO(4)-promoted oxidative ring opening of the resulting diols 2, followed by an intramolecular Knoevenagel condensation, furnished versatile dihydrotropones 6. Maintaining Z-configuration in the
Strahil Berkov et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 59(3-4), 184-186 (2004-07-10)
The alkaloid spectra of Datura innoxia plants grown in Egypt and Bulgaria were investigated by GC-MS. Thirty-eight alkaloids were detected in the roots, leaves and fruits of the plants. Five new alkaloids for D. innoxia are reported. Alkaloid spectra of
Honglei Liu et al.
Journal of the American Chemical Society, 136(6), 2625-2629 (2014-01-24)
The first metal-catalyzed [6 + 3] cycloaddition of tropone with azomethine ylides has been developed. With the use of a chiral ferrocenylphosphine-copper(I) complex as the catalyst, the asymmetric variant of the [6 + 3] cycloaddition has also been successfully achieved.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service