Skip to Content
Merck
All Photos(2)

Key Documents

247065

Sigma-Aldrich

(S)-cis-Verbenol

95%

Synonym(s):

(1S,2S)-4,6,6-Trimethylbicyclo[3.1.1]hept-3-en-2-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
2206582
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

optical activity

[α]20/D −9° in chloroform
[α]20/D +10° in ethanol

optical purity

ee: ≥50% (GLC)

mp

62-65 °C (lit.)

SMILES string

CC1=C[C@H](O)[C@H]2C[C@@H]1C2(C)C

InChI

1S/C10H16O/c1-6-4-9(11)8-5-7(6)10(8,2)3/h4,7-9,11H,5H2,1-3H3/t7-,8+,9-/m0/s1

InChI key

WONIGEXYPVIKFS-YIZRAAEISA-N

Looking for similar products? Visit Product Comparison Guide

General description

(S)-cis-verbenol is an aggregation pheromone component in the bark beetles. It shows potent anti-oxidative and anti-inflammatory activities.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

P J Rice et al.
Journal of economic entomology, 87(5), 1172-1179 (1994-10-01)
House flies, Musca domestica (L.), and their eggs were treated with 22 monoterpenoids to determine the topical, fumigant, and ovicidal activity of each compound. Fumigant activity of 14 monoterpenoids were examined further using red flour beetles, Tribolium castaneum (Herbst). Third-instar
Interconversion of verbenols and verbenone by identified yeasts isolated from the spruce bark beetle Ips typographus.
Leufven A, et al.
Journal of Chemical Ecology, 10(9), 1349-1361 (1984)
Jeremy D Allison et al.
Journal of economic entomology, 105(3), 923-929 (2012-07-21)
Stressed or damaged pine (Pinus sp.) trees in the southeastern United States are often colonized simultaneously by three southern Ips species (Coleoptera: Curculionidae: Scolytinae): small southern pine engraver, Ips avulsus (Eichhoff); sixspined ips, Ips calligraphus (Germar); and eastern fivespined ips
Martin N Andersson et al.
Journal of chemical ecology, 37(8), 899-911 (2011-07-14)
Orientation for insects in olfactory landscapes with high semiochemical diversity may be a challenging task. The partitioning of odor plumes into filaments that are interspersed with pockets of 'clean air' may help filament discrimination and upwind flight to attractive sources
Kairomone response in Thanasimus predators to pheromone components of Ips typographus.
Bakke A and Kvamme T.
Journal of Chemical Ecology, 7(2), 305-312 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service