Skip to Content
Merck
All Photos(1)

Key Documents

246344

Sigma-Aldrich

9-Acridinecarboxylic acid hydrate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9NO2 · xH2O
CAS Number:
Molecular Weight:
223.23 (anhydrous basis)
Beilstein:
171327
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

290 °C (dec.) (lit.)

functional group

carboxylic acid

SMILES string

[H]O[H].OC(=O)c1c2ccccc2nc3ccccc13

InChI

1S/C14H9NO2.H2O/c16-14(17)13-9-5-1-3-7-11(9)15-12-8-4-2-6-10(12)13;/h1-8H,(H,16,17);1H2

InChI key

ZQAZWHSZYVXGMP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

9-Acridinecarboxylic acid hydrate has been used to synthesize a DNA intercalator in order to capture double-stranded DNAs (dsDNAs) duplex from the solution.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrication of DNA Nanowires by Orthogonal Self-Assembly and DNA Intercalation on a Au Patterned Si/SiO2 Surface.
Kobayashi K, et al.
Langmuir, 24(22), 13203-13211 (2008)
Synthesis and chemiluminescent properties of novel biotinylated acridinium esters.
Agiamarnioti k, et al.
Acta Chimica Slovenica, 51(1), 67-76 (2004)
Wolf-Ulrich Palm
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 17(7), 964-974 (2018-06-20)
Dark and photolysis reactions in solution were investigated for 9-acridinecarboxaldehyde (ACL). ACL reacts in the dark at T = 20 °C and pH = 7.0 in an air saturated solution to the main product 9-acridinecarboxylic acid (ACA) and to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service