237647
L−(+)-α-Phenylglycine
99%
Synonym(s):
(S)-(+)-2-Phenylglycine, S-(+)-α-Aminophenylacetic acid, L-2-Phenylglycine
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
C6H5CH(NH2)CO2H
CAS Number:
Molecular Weight:
151.16
Beilstein:
2208675
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
form
powder
optical activity
[α]20/D +155°, c = 1 in 1 M HCl
reaction suitability
reaction type: solution phase peptide synthesis
mp
>300 °C (lit.)
application(s)
peptide synthesis
SMILES string
N[C@H](C(O)=O)c1ccccc1
InChI
1S/C8H9NO2/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7H,9H2,(H,10,11)/t7-/m0/s1
InChI key
ZGUNAGUHMKGQNY-ZETCQYMHSA-N
Related Categories
Application
Chiral starting material.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Henry Kurniawan et al.
Cell metabolism, 31(5), 920-936 (2020-03-28)
Regulatory T cells (Tregs) maintain immune homeostasis and prevent autoimmunity. Serine stimulates glutathione (GSH) synthesis and feeds into the one-carbon metabolic network (1CMet) essential for effector T cell (Teff) responses. However, serine's functions, linkage to GSH, and role in stress responses in
James J Lynch et al.
Pain, 125(1-2), 136-142 (2006-06-20)
Gabapentin and pregabalin have been demonstrated, both in animal pain models and clinically, to be effective analgesics particularly for the treatment of neuropathic pain. The precise mechanism of action for these two drugs is unknown, but they are generally believed
José Alixandre de Sousa Luis et al.
Molecules (Basel, Switzerland), 15(1), 128-137 (2010-01-30)
Hydantoins and their derivatives constitute a group of pharmaceutical compounds with anticonvulsant and antiarrhythmic properties, and are also used against diabetes. N-3 and C-5 substituted imidazolidines are examples of such products. As such, we have developed a synthesis of 2,4-dione
Shohei Tashiro et al.
Inorganic chemistry, 50(1), 4-6 (2010-12-01)
The optically active cobalt(III) complex with chiral cyclen, (2S,5S,8S,11S)-2,5,8,11-tetraethyl-1,4,7,10-tetraazacyclododecane, preferentially binds to D-phenylglycine (D-Phg) or D-t-leucine (D-t-Leu) rather than L-Phg or L-t-Leu, respectively, with 20% de in dimethyl sulfoxide at 293 K. Comparative studies on the crystal structures of cobalt(III)
Hyung Min Kim et al.
The Journal of chemical physics, 128(18), 184313-184313 (2008-06-06)
We investigated the conformational structures of L-phenylglycine in the gas phase by photoionization and double resonance spectroscopy techniques as well as high-level ab initio calculations. The UV-UV and IR-UV double resonance spectroscopy suggested that there exists only one conformer that
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service