23265
N-Chlorobenzenesulfonamide sodium salt
~28% active chlorine basis
Synonym(s):
Benzene chloramine, Chloramine B, Sodium (phenylsulfonyl)chloramide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5SO2NClNa · aq
CAS Number:
Molecular Weight:
213.62 (anhydrous basis)
Beilstein:
3599287
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
form
solid
reaction suitability
reagent type: oxidant
concentration
~28% (active chlorine)
SMILES string
O.[Na]N(Cl)S(=O)(=O)c1ccccc1
InChI
1S/C6H5ClNO2S.Na.H2O/c7-8-11(9,10)6-4-2-1-3-5-6;;/h1-5H;;1H2/q-1;+1;
InChI key
YVICIJMKLANTNN-UHFFFAOYSA-N
Related Categories
Application
Used in investigations of the toxicity response of electroactive microbial biofilms
Catalyst for rearrangement of aziridinofullerenes to azafulleroids
Oxidizing agent for polymerization of thiophenol, synthesis of o-aminobenzenesulfonic acids and ciproflaxin
Decontaminant for mustard
Catalyst for rearrangement of aziridinofullerenes to azafulleroids
Oxidizing agent for polymerization of thiophenol, synthesis of o-aminobenzenesulfonic acids and ciproflaxin
Decontaminant for mustard
Other Notes
Review
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Corr. 1B
Supplementary Hazards
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
E George et al.
Acta crystallographica. Section C, Crystal structure communications, 56 ( Pt 10), 1208-1209 (2000-10-12)
In the title compound, sodium N-chlorobenzenesulfonamide sesquihydrate, Na(+).C(6)H(5)ClNO(2)S(-).1.5H(2)O, the sodium ion exhibits octahedral coordination by O atoms from three water molecules and by three sulfonyl O atoms of three different N-chlorobenzenesulfonamide anions. A two-dimensional polymeric layer consists of units, each
[Comparative sensitivity of microorganisms to monochloramine B].
T B Krucheniuk
Gigiena i sanitariia, (6)(6), 75-77 (1985-06-01)
[Quantitative evaluation of comparative resistance of various phages and viruses to monochloramine-B].
P P Liarskiĭ et al.
Gigiena i sanitariia, (3)(3), 22-25 (1990-03-01)
G V Lobacheva et al.
Kosmicheskaia biologiia i aviakosmicheskaia meditsina, 19(1), 70-72 (1985-01-01)
The comparative toxicity of halogen-containing oxidizing agents was investigated with the purpose of their utilization as urine preservatives in water reclamation systems. It was found that the high toxicity of the agents (LD50 of agent 1 was 15.7 +/- 1.1
[Chemical method of sterilizing plastic bacteriological dishes].
B E Bilich et al.
Laboratornoe delo, (3)(3), 176-178 (1984-01-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service