Skip to Content
Merck
All Photos(1)

Key Documents

227757

Sigma-Aldrich

Palladium

sulfided, extent of labeling: 5 wt. % loading (dry basis), matrix activated carbon, wet support, (wet)

Synonym(s):

Palladium on carbon, Pd/C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd
Molecular Weight:
106.42
MDL number:
UNSPSC Code:
11101718
PubChem Substance ID:
NACRES:
NA.22

Quality Level

form

(wet)

contains

≤50% water

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

extent of labeling

5 wt. % loading (dry basis)

weight

Unit weight excludes weight of water

matrix

activated carbon, wet support

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Stille reaction
  • Hydrogenation reactions
  • Suzuki-Miyaura cross-coupling and Heck-Mizoroki reactions
  • Deoxygenation
  • Oxidation reactions
  • Coupling reactions

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John R Hornick et al.
Journal of experimental & clinical cancer research : CR, 31, 41-41 (2012-05-04)
Sigma-2 receptor ligands have been studied for treatment of pancreatic cancer because they are preferentially internalized by proliferating cells and induce apoptosis. This mechanism of apoptosis is poorly understood, with varying reports of caspase-3 dependence. We evaluated multiple sigma-2 receptor
Mauro Niso et al.
Chembiochem : a European journal of chemical biology, 16(7), 1078-1083 (2015-03-11)
Although sigma-2 (σ2 ) receptors are still enigmatic proteins, they are promising targets for tumor treatment and diagnosis. With the aim of clarifying their role in oncology, we developed a σ2 -selective fluorescent tracer (compound 5) as a specific tool
Carmen Abate et al.
ChemMedChem, 5(2), 268-273 (2010-01-16)
Sigma-2 (sigma(2)) binding sites are an emerging target for anti-neoplastic agents due to the strong apoptotic effect exhibited by sigma(2) agonists in vitro and the overexpression of these sites in tumor cells. Nonetheless, no sigma(2) receptor protein has been identified.
Carmen Abate et al.
ChemMedChem, 6(1), 73-80 (2010-11-12)
Many new chemotherapeutic agents are under preclinical investigation and, despite efforts to more selectively target cancer cells, limitations such as toxicity and inherent resistance are often encountered. Therefore, alternative strategies are needed to treat cancer and overcome such limitations. We
Mauro Niso et al.
ChemMedChem, 8(12), 2026-2035 (2013-10-10)
With the aim of contributing to the development of novel antitumor agents, high-affinity σ2 receptor agonists were developed, with 6,7-dimethoxy-2-[4-[1-(4-fluorophenyl)-1H-indol-3-yl]butyl]-1,2,3,4-tetrahydroisoquinoline (15) and 9-[4-(6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-2-yl)butyl]-9H-carbazole (25) showing exceptional selectivity for the σ2 subtype. Most of the compounds displayed notable antiproliferative activity in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service