Skip to Content
Merck
All Photos(2)

Key Documents

227609

Sigma-Aldrich

Decaborane(14)

white

Synonym(s):

Boron hydride, Decaborane, Decaboron tetradecahydride, NSC 39828, Tetradecahydrodecaborane, nido-Decaborane(14)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
B10H14
CAS Number:
Molecular Weight:
122.22
EC Number:
MDL number:
UNSPSC Code:
12352000

vapor pressure

0.15 mmHg ( 20 °C)

Quality Level

form

solid

reaction suitability

reagent type: reductant

color

white

bp

213 °C (lit.)

mp

98-100 °C (lit.)

density

0.94 g/mL at 25 °C (lit.)

SMILES string

[BH]4321[BH]765[BH]984[BH]%11%103[BH]%13%122[BH]%1471[BH]%156([BH]%1695[BH]%118([H]%10)[H]%16)[BH]%14%13([H]%12)[H]%15

InChI

1S/B10H14/c11-5-1-2-3(1,5)7(2,9(3,5,11)13-7)8(2)4(1,2)6(1,5)10(4,8,12-6)14-8/h1-10H

InChI key

XAMMYYSPUSIWAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Boron source for ion-implantation processes.
Reactant for:
  • Chemical vapor deposition of boron nitride nanosheets on Ni or Cu foils in the presence of ammonia
  • Stereoselective catalytic hydrogenation reactions
  • Chemical hydrogen storage
  • Preparation of carborane-conjugated quinoline carboxamide ligands of translocator protein for boron neutron capture therapy (BNCT)
  • Synthesis of hydrogenated boron clusters B12Hn with controlled hydrogen content
  • Preparation of alkenyldecaboranes by regioselective transition-metal-catalyzed decaborane-alkyne hydroboration reactions

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhu Yinghuai et al.
Journal of the American Chemical Society, 129(20), 6507-6512 (2007-05-03)
Well dispersed ruthenium(0) nanoparticles, stabilized in the ionic liquid agent, trihexyltetradecylphosphonium dodecylbenzenesulfonate, have been successfully prepared via a reduction reaction of the precursor [CpRuCp*RuCp*]PF6 (Cp* = C5Me5). The ruthenium(0) nanoparticles were shown to catalyze the isotope exchange reaction between 10B
Kiminori Ohta et al.
Bioorganic & medicinal chemistry letters, 14(23), 5913-5918 (2004-10-27)
We designed and synthesized novel retinoid X receptor (RXR)-selective antagonists bearing a carborane moiety. Compounds 8a-d or 9a-d themselves have no differentiation-inducing activity toward HL-60 cells and no inhibitory activity towards a retinoic acid receptor (RAR) agonist. However, they inhibit
Ilias Sioutis et al.
The journal of physical chemistry. A, 110(45), 12528-12534 (2006-11-10)
The interaction of decaborane (B(10)H(14)) with the I(-) ion and the (isoelectronic) Xe atom is investigated using a number of theoretical methods: MP2, CCSD(T), CCSD, spin-orbit CISD, and DFT using the B3LYP, B3PW91, PW91PW91, and PBE0 methods. All non-DFT and
Yuqi Li et al.
Inorganic chemistry, 47(20), 9193-9202 (2008-09-20)
Unlike in conventional organic solvents, where Lewis base catalysts are required, decaborane dehydrogenative alkyne-insertion reactions proceed rapidly in biphasic ionic-liquid/toluene mixtures with a wide variety of terminal and internal alkynes, thus providing efficient, one-step routes to functional o-carborane 1-R-1,2-C2B10H11 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service