Skip to Content
Merck
All Photos(1)

Key Documents

225827

Sigma-Aldrich

N,N′-Disuccinimidyl carbonate

≥95%, for peptide synthesis

Synonym(s):

N-Succinimidyl carbonate, DSC, Di(N-succinimidyl) carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8N2O7
CAS Number:
Molecular Weight:
256.17
Beilstein:
1499137
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Product Name

N,N′-Disuccinimidyl carbonate, ≥95%

Assay

≥95%

form

powder or crystals

reaction suitability

reaction type: Carbonylations

mp

190 °C (dec.) (lit.)

application(s)

peptide synthesis

functional group

imide

storage temp.

2-8°C

SMILES string

O=C1CCC(=O)N1OC(=O)ON2C(=O)CCC2=O

InChI

1S/C9H8N2O7/c12-5-1-2-6(13)10(5)17-9(16)18-11-7(14)3-4-8(11)15/h1-4H2

InChI key

PFYXSUNOLOJMDX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N′-Disuccinimidyl carbonate is used for the HPLC determination of amino compounds.

Application

N,N′-Disuccinimidyl carbonate may be used:
  • As an activating reagent for the reactive patterning of polymer brushes to couple primary amines via post-functionalization.
  • As an alkoxycarbonylating reagent for a variety of primary and sterically hindered secondary alcohols, which further allows the synthesis of different structurally diverse carbamates.
  • As a coupling agent for the synthesis of thiophospholipids, commonly used for anchoring biomembranes to gold surfaces.
It can also be used to prepare a heterobifunctional, cleavable linker-SVEC-(succinimide vinylsufonylethyl carbonate)-for thiol modified-DNA.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT RE 2 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactive patterning via post-functionalization of polymer brushes utilizing disuccinimidyl carbonate activation to couple primary amines
Diamanti S, et al.
Polymer, 49(17), 3770-3779 (2008)
Chang-Juan Tao et al.
Radiotherapy and oncology : journal of the European Society for Therapeutic Radiology and Oncology, 115(3), 407-411 (2015-05-31)
To assess whether consensus guideline-based atlas-based auto-segmentation (ABAS) reduces interobserver variation and improves dosimetric parameter consistency for organs at risk (OARs) in nasopharyngeal carcinoma (NPC). Eight radiation oncologists from 8 institutes contoured 20 OARs on planning CT images of 16
N, N?-Disuccinimidyl Carbonate: A Useful Reagent for Alkoxycarbonylation of Amines
Ghosh KA, et al.
Tetrahedron Letters, 33(20), 2781-2781 (1992)
Tetrahedron Letters, 33, 2781-2781 (1992)
N,N?-Disuccinimidyl carbonate as a coupling agent in the synthesis of thiophospholipids used for anchoring biomembranes to gold surfaces
Boden N, et al.
Tetrahedron, 54(38), 11537-11548 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service