Skip to Content
Merck
All Photos(1)

Key Documents

22390

Sigma-Aldrich

Cerium(IV) oxide

greener alternative

≥99.0%

Synonym(s):

Ceric oxide, Ceric oxide, ceria

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CeO2
CAS Number:
Molecular Weight:
172.11
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0%

form

solid

reaction suitability

core: cerium
reagent type: catalyst

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

loss

≤0.5% loss on ignition

density

7.13 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

O=[Ce]=O

InChI

1S/Ce.2O

InChI key

CETPSERCERDGAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and is intended for Molecular Solar Thermal Energy Storage Systems (MOST). Click here for more information.

Application

The additon of CeO2 to CuO/TiO2 increases specific surface area and inhibits sintering and pore size growth at high temperatures.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dengsong Zhang et al.
Nanoscale, 5(3), 1127-1136 (2013-01-04)
The MnO(x) and CeO(x) were in situ supported on carbon nanotubes (CNTs) by a poly(sodium 4-styrenesulfonate) assisted reflux route for the low-temperature selective catalytic reduction (SCR) of NO with NH(3). X-Ray diffraction (XRD), transmission electron microscopy (TEM), high-resolution TEM (HRTEM)
Susmita Bandyopadhyay et al.
Journal of hazardous materials, 241-242, 379-386 (2012-10-23)
Cerium oxide (CeO(2)) and zinc oxide (ZnO) nanoparticles (NPs) are extensively used in a variety of instruments and consumer goods. These NPs are of great concern because of potential toxicity towards human health and the environment. The present work aimed
Gao Li et al.
Chemical communications (Cambridge, England), 48(98), 12005-12007 (2012-11-07)
The Au(25)(SR)(18)/CeO(2) nanocluster catalyst showed high activity in the homocoupling of aryl iodides (e.g. up to 99.8% yield with iodobenzene) and excellent recyclability.
Peng Zhang et al.
ACS nano, 6(11), 9943-9950 (2012-10-27)
Biotransformation is a critical factor that may modify the toxicity, behavior, and fate of engineered nanoparticles in the environment. CeO(2) nanoparticles (NPs) are generally recognized as stable under environmental and biological conditions. The present study aims to investigate the biotransformation
Lijuan Zhao et al.
ACS nano, 6(11), 9615-9622 (2012-10-12)
The rapid development of nanotechnology will inevitably release nanoparticles (NPs) into the environment with unidentified consequences. In addition, the potential toxicity of CeO(2) NPs to plants and the possible transfer into the food chain are still unknown. Corn plants (Zea

Articles

As with all types of fuel cells, a Solid Oxide Fuel Cell (SOFC) is capable of efficiently transforming chemical energy into electrical energy.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service