Skip to Content
Merck
All Photos(1)

Key Documents

218243

Sigma-Aldrich

(1R)-(−)-Myrtenal

98%

Synonym(s):

(−)-Myrtenal, (1R)-2-Pinen-10-al, (1R)-6,6-Dimethylbicyclo[3.1.1]hept-2-en-2-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
Beilstein:
2961587
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

optical activity

[α]22/D −15°, neat

refractive index

n20/D 1.503 (lit.)

bp

220-221 °C (lit.)

density

0.987 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)C1=CCC2CC1C2(C)C

InChI

1S/C10H14O/c1-10(2)8-4-3-7(6-11)9(10)5-8/h3,6,8-9H,4-5H2,1-2H3/t8-,9-/m0/s1

InChI key

KMRMUZKLFIEVAO-IUCAKERBSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

172.4 °F

Flash Point(C)

78 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service