21190
Calcium lactobionate monohydrate
≥98.0% (KT)
Synonym(s):
Lactobionic acid monohydrate
About This Item
Recommended Products
Assay
≥98.0% (KT)
form
solid
optical activity
[α]20/D +23.5±1°, c = 3% in H2O
impurities
2.5-6.5% water
color
colorless
mp
190 °C ((374 °F ))
SMILES string
O.O.OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C(=O)O[Ca]OC(=O)[C@H](O)[C@@H](O)[C@H](O[C@@H]2O[C@H](CO)[C@H](O)[C@H](O)[C@H]2O)[C@H](O)CO
InChI
1S/2C12H22O12.Ca.2H2O/c2*13-1-3(15)10(7(18)8(19)11(21)22)24-12-9(20)6(17)5(16)4(2-14)23-12;;;/h2*3-10,12-20H,1-2H2,(H,21,22);;2*1H2/q;;+2;;/p-2/t2*3-,4-,5+,6+,7-,8-,9-,10-,12+;;;/m11.../s1
InChI key
FALNADDKNDADFQ-WNSOHMSISA-L
Looking for similar products? Visit Product Comparison Guide
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service