Skip to Content
Merck
All Photos(4)

Key Documents

208833

Sigma-Aldrich

Ruthenium(IV) oxide hydrate

powder

Synonym(s):

Hydrous ruthenium oxide, Ruthenium dioxide hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
RuO2 · xH2O
CAS Number:
Molecular Weight:
133.07 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

form

powder

reaction suitability

reagent type: catalyst
core: ruthenium

SMILES string

O.O=[Ru]=O

InChI

1S/H2O.2O.Ru/h1H2;;;

InChI key

FGEKTVAHFDQHBU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beena K Balan et al.
Inorganic chemistry, 51(18), 9766-9774 (2012-09-06)
Chemical-functionalization-induced switching in the property of a hybrid system composed of a hollow carbon nanofiber (CNF) and Pt and RuO(2) nanoparticles from charge storage to electrocatalysis is presented. The results of this study show how important it is to have
Wen-Yan Liang et al.
Huan jing ke xue= Huanjing kexue, 31(6), 1464-1469 (2010-08-12)
The paper studied the variation of algal activity during the electrochemical inactivation and the influence factors by the use of TTC-dehydrogenase activity and neutral red staining assays. The treatment reactor was consisted of Ti/RuO2 rod as anode and stainless steel
Chengzhi Hu et al.
Journal of environmental sciences (China), 24(2), 297-302 (2012-06-05)
A polyaluminum containing a high concentration of Al13 polymer and active chlorine (PACC) was successfully synthesized by a new electrochemical reactor using Ti/RuO2-TiO2 anodes. PACC can potentially be used as a dual-function chemical reagent for water treatment. The obtained results
Guolei Xiang et al.
Scientific reports, 2, 801-801 (2012-11-13)
Controls over the atomic dispersity and particle shape of noble metal catalysts are the major qualities determining their usability in industrial runs, but they are usually difficult to be simultaneously realized. Inspired from the Deacon catalyst in which RuO(2) can
Ramin M A Tehrani et al.
Biosensors & bioelectronics, 38(1), 278-283 (2012-06-30)
A non-enzymatic glucose sensor of multi-walled carbon nanotube-ruthenium oxide/composite paste electrode (MWCNT-RuO(2)/CPE) was developed. The electrode was characterized by using XRD, SEM, TEM and EIS. Meanwhile, cyclic voltammetry and amperometry were used to check on the performances of the MWCNT-RuO(2)/CPE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service