Skip to Content
Merck
All Photos(1)

Documents

206326

Sigma-Aldrich

Dibromochloromethane

97%

Synonym(s):

Chlorodibromomethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br2CHCl
CAS Number:
Molecular Weight:
208.28
Beilstein:
1731046
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.547 (lit.)

bp

119-120 °C/748 mmHg (lit.)

mp

−22 °C (lit.)

solubility

acetone: soluble
diethyl ether: soluble
ethanol: soluble

density

2.451 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ClC(Br)Br

InChI

1S/CHBr2Cl/c2-1(3)4/h1H

InChI key

GATVIKZLVQHOMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Affords bromochlorocarbene under phase-transfer conditions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis, 857-857 (1991)
Qiang Zeng et al.
Environment international, 54, 134-140 (2013-03-05)
Toxicological studies showed that trihalomethanes (THMs), the most abundant classes of disinfection by-products (DBPs) in drinking water, impaired male reproductive health, but epidemiological evidence is limited and inconsistent. This study aimed to examine the associations of baseline blood THMs with
M N Kostopoulou et al.
Chemosphere, 40(5), 527-532 (2000-02-09)
An investigation into the occurrence of volatile organic compounds (VOCs) was conducted for a period of two years in the surface waters of Northern Greece. Samples from four rivers and five lakes were taken seasonally and analyzed for VOCs. The
M Luciene da Silva et al.
Toxicology letters, 106(1), 49-57 (1999-06-23)
Trihalomethanes (THMs; chloroform, bromoform, bromodichloromethane, dibromochloromethane), formed as by-products of chlorine disinfection, are found to occur in combination in drinking water supplies. THMs are metabolized by cytochromes P-450 and are likely substrates of CYP2E1. Therefore, it is possible that mixed
S Landi et al.
Mutagenesis, 14(5), 479-482 (1999-09-03)
The brominated trihalomethanes (THMs) are mutagenic and carcinogenic disinfection by-products frequently found in chlorinated drinking water. They can be activated to mutagens by the product of the glutathione S-transferase-Theta (GSTT1++-1) gene in Salmonella RSJ100, which has been transfected with this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service