Skip to Content
Merck
All Photos(2)

Documents

205311

Sigma-Aldrich

Lead(II) perchlorate hydrate

98%

Synonym(s):

Lead perchlorate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(ClO4)2·xH2O
CAS Number:
Molecular Weight:
406.10 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

reaction suitability

reagent type: oxidant

density

2.6 g/mL at 25 °C (lit.)

SMILES string

O.[PbH2++].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O

InChI

1S/2ClHO4.H2O.Pb/c2*2-1(3,4)5;;/h2*(H,2,3,4,5);1H2;/q;;;+2/p-2

InChI key

ZIJXNFOWEHUQHM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

  • Reactant for preparation of lead coordination polymers

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Ox. Sol. 2 - Repr. 1A - STOT RE 2

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service