202312
Poly(propylene glycol)
average Mn ~725
Synonym(s):
PPG, Poly(propylene oxide)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
H[OCH(CH3)CH2]nOH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
vapor pressure:
<0.01 mmHg ( 20 °C)
Recommended Products
vapor density
>1 (vs air)
vapor pressure
<0.01 mmHg ( 20 °C)
mol wt
average Mn ~725
contains
130-190 ppm proprietary phenolic antioxidant
refractive index
n20/D 1.449
viscosity
115 cSt(25 °C)(lit.)
hydroxyl value
147 mg KOH/g
solubility
water: miscible (completely)
density
1.007 g/mL at 25 °C
SMILES string
CC(O)CO
InChI
1S/C6H14O3/c1-5(8)4-9-6(2)3-7/h5-8H,3-4H2,1-2H3
InChI key
DUFKCOQISQKSAV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Recycling of waste materials: Utilization of propylene glycol in the recycling of waste poly(vinyl chloride) to enhance the mechanical properties of new polymer composites (Hilary et al., 2021).
- Dental composite improvement: Enhancement of dental composites′ conversion rates through the use of poly(propylene glycol) and urethane dimethacrylates, while also examining cytocompatibility (Walters et al., 2016).
- 3D printing of scaffolds: Use of poly(propylene fumarate) and propylene glycol in 3D printing of resorbable tissue engineering scaffolds, demonstrating innovation in medical applications (Childers et al., 2015).
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
445.0 °F - closed cup
Flash Point(C)
229.44 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
R De Lisi et al.
Physical chemistry chemical physics : PCCP, 13(27), 12571-12577 (2011-06-15)
The study highlighted the main forces driving the formation of hydroxypropyl-cyclodextrins (HP-CDs) + poly(propylene) glycol 725 g mol(-1) inclusion complexes. The temperature parameter was chosen as the variable to modulate the hydrophobicity of the polymer, and consequently ITC experiments as
Izzuddin Zaman et al.
Nanoscale, 4(15), 4578-4586 (2012-06-19)
In spite of extensive studies conducted on carbon nanotubes and silicate layers for their polymer-based nanocomposites, the rise of graphene now provides a more promising candidate due to its exceptionally high mechanical performance and electrical and thermal conductivities. The present
Mohammed Taghi Zafarani-Moattar et al.
Biotechnology progress, 28(1), 146-156 (2011-09-29)
In this work, we proposed a novel aqueous biphasic system (ABS) composed of polypropylene glycol P400 (PPG P400) and hydrophilic ionic liquids (IL), 1-alkyl-3-methylimidazolium bromide (alkyl = ethyl or butyl), forming an upper polymer-rich phase and a lower IL-rich phase
Flávia Chiva Carvalho et al.
Journal of biomedical nanotechnology, 8(2), 280-289 (2012-04-21)
In the last few decades, nanotechnology has led to an advance in the development of topical drug delivery. Nanostructured drug delivery systems enable the compartmentalization of drugs in restricted environments, modifying the release profile and maintaining the required drug concentration
F Palazzi et al.
International endodontic journal, 45(2), 129-135 (2011-09-13)
To investigate the surface tension characteristics of 5.25% sodium hypochlorite and three recently introduced sodium hypochlorite solutions, which had been modified to reduce their surface tension: Chlor-Xtra, Hypoclean A and Hypoclean B. Freshly produced MilliQ water was used as a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service