Skip to Content
Merck
All Photos(1)

Documents

197327

Sigma-Aldrich

S-Acetylmercaptosuccinic anhydride

96%

Synonym(s):

2-(Acetylthio)succinic anhydride, SAMSA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O4S
CAS Number:
Molecular Weight:
174.17
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

96%

mp

83-86 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)SC1CC(=O)OC1=O

InChI

1S/C6H6O4S/c1-3(7)11-4-2-5(8)10-6(4)9/h4H,2H2,1H3

InChI key

AHTFMWCHTGEJHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

S-Acetylmercaptosuccinic anhydride is an amine reactive reagent, containing a sulfhydryl group. The anhydride region opens up, when attacked by an amine nucleophile, forming amine linkage. However, during this ring opening reaction, a free carboxylate group is formed rendering the molecule negatively charged. Activity to proteins and conformation of the molecule us affected by the charge reversal.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Introduction of sulfhydryl groups into proteins using acetylmercaptosuccinic anhydride
Klotz IM, et al.
Archives of Biochemistry and Biophysics, 96(3), 605-612 (1932)
L Greenfield et al.
Bioconjugate chemistry, 1(6), 400-410 (1990-11-01)
Ricin A chain immunotoxins disulfide cross-linked with conventional, sterically unhindered reagents have unsatisfactorily short circulating life times in vivo. (Acetylthio)succinic anhydride, a thiolating reagent with partial steric hindrance of the sulfur atom, does not remedy this situation. Sulfosuccinimidyl N-[3-(acetylthio)-3-methylbutyryl]-beta- alaninate
Monica A Serban et al.
Biomacromolecules, 8(9), 2821-2828 (2007-08-19)
Hyaluronan (HA) derivatives containing thiol-reactive electrophilic esters were prepared to react with thiol-modified macromolecules to give cross-linker-free hydrogels. Specifically, HA was converted to two haloacetate derivatives, HA bromoacetate (HABA) and HA iodoacetate (HAIA). In cytotoxicity assays, these reactive macromolecules predictably
Fabio Cannone et al.
The journal of physical chemistry. B, 110(33), 16491-16498 (2006-08-18)
A fluorescein derivative (SAMSA) bound to gold nanoparticles of different diameters is investigated by time-resolved fluorescence at the single molecule level in a wide dynamic range, from nanosecond to second time scale. The significant decrease of both SAMSA excited state
Hermanson GT et al.
Bioconjugate Techniques null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service