Skip to Content
Merck
All Photos(1)

Key Documents

194751

Sigma-Aldrich

N-tert-Butylhydroxylamine hydrochloride

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CNHOH · HCl
CAS Number:
Molecular Weight:
125.60
Beilstein:
3546053
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

solid

mp

183-185 °C (lit.)

SMILES string

Cl.CC(C)(C)NO

InChI

1S/C4H11NO.ClH/c1-4(2,3)5-6;/h5-6H,1-3H3;1H

InChI key

DCSATTBHEMKGIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-tert-Butylhydroxylamine hydrochloride was used in spin trapping of short-lived radicals. It was also used in the synthesis of α-ketoamides and 3-spirocyclopropanated 2-azetidinones.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyun Jeong Kim et al.
Redox report : communications in free radical research, 10(6), 287-293 (2006-01-28)
Heat shock may increase oxidative stress due to increased production of reactive oxygen species and/or the promotion of cellular oxidation events. Therefore, compounds that scavenge reactive oxygen species may regulate heat shock-induced cell death. Recently, it has been shown that
Yu-Kyung Kim et al.
Clinical hemorheology and microcirculation, 40(4), 315-324 (2009-01-08)
Irradiation has been shown to induce biochemical changes in stored red blood cells (RBCs) and to generate reactive oxygen species (ROS). This study evaluated the hemorheological properties, the degree of lipid peroxidation and the oxidative susceptibility of irradiated RBCs. Furthermore
A New Three-Component Cascade Reaction to Yield 3-Spirocyclopropanated?-Lactams.
Zanobini A, et al.
European Journal of Organic Chemistry, 2006(5), 1251-1255 (2006)
C Lagercrantz
Free radical research communications, 14(5-6), 395-407 (1991-01-01)
Spin trapping of short-lived R. radicals is done by use of N-tert-butylhydroxylamine (1) and H2O2. The hydroxylamine is oxidized to the radical t-BuN(O)H (2) which is converted into the spin trap 2-methyl-2-nitrosopropane (3). Simultaneously, hydroxyl radicals .OH are formed from
Ludmila A Voloboueva et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 21(14), 4077-4086 (2007-07-28)
Age-related macular degeneration (AMD) is the leading cause of severe visual impairment in the elderly in developed countries. AMD patients have elevated levels of iron within the retinal pigment epithelia (RPE), which may lead to oxidative damage to mitochondria, disruption

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service