Skip to Content
Merck
All Photos(2)

Key Documents

191094

Sigma-Aldrich

Poly(oxy-1,4-phenylenesulfonyl-1,4-phenylene)

pellets

Synonym(s):

Poly(1,4-phenylene ether-sulfone)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H8O3S)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

pellets

Quality Level

transition temp

Tm (DSC) 213 °C (onset)

density

1.37 g/mL at 25 °C (lit.)

SMILES string

Oc1ccc(cc1)S(=O)(=O)c2ccc(O)cc2

InChI key

VPWNQTHUCYMVMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mario Bonomini et al.
Therapeutics and clinical risk management, 14, 1-9 (2018-01-04)
The success and the quality of hemodialysis therapy are mainly related to both clearance and biocompatibility properties of the artificial membrane packed in the hemodialyzer. Performance of a membrane is strongly influenced by its interaction with the plasma protein repertoire
Daniel Schneditz et al.
Clinical hemorheology and microcirculation, 58(3), 455-469 (2013-11-21)
It was the aim to examine the fluid flow in blood and dialysate compartments of highly permeable hollow fiber dialyzers where internal filtration contributes to solute removal but where excessive filtration bears a risk of cell activation and damage. Flow
Shogo Takamuku et al.
ChemSusChem, 6(2), 308-319 (2013-01-12)
The morphologies and proton-transport efficiencies of segmented copoly(arylene ether sulfone) ionomers that contain tetrasulfonated sequences are compared with the corresponding copolymers with disulfonated sequences. Tetrasulfonated 4,4'-bis[(4-chlorophenyl)sulfonyl]-1,1'-biphenyl (sBCPSBP) is synthesized by metalation and sulfination. This new monomer is then used in
Eugenia Villaverde-de-Sáa et al.
Analytica chimica acta, 740, 50-57 (2012-07-31)
A method for the determination of seven perfluorinated carboxylic acids and perfluorooctane sulphonate (PFOS) in aqueous samples using low-cost polymeric sorptive extraction as sample preparation technique, followed by liquid chromatography-tandem mass spectrometry (LC-MS/MS) determination has been developed and validated. Simplicity
Dongsheng Wang et al.
Langmuir : the ACS journal of surfaces and colloids, 28(37), 13284-13293 (2012-08-17)
In the present study, photoresponsive surface molecularly imprinted poly(ether sulfone) microfibers are prepared via nitration reaction, the wet-spinning technique, surface nitro reduction reaction, and surface diazotation reaction for the selectively photoregulated uptake and release of 4-hydrobenzoic acid. The prepared molecularly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service