Skip to Content
Merck
All Photos(2)

Documents

181161

Sigma-Aldrich

Nylon 12

pellets

Synonym(s):

Polylauryllactam

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H23NO)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

pellets

Quality Level

particle size

3-4 mm

transition temp

Tm 178 °C
Tg 37 °C

density

1.01 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCCCCCCCCCN1

Application

  • Replicating and Redesigning Ankle-Foot Orthoses with 3D Printing for Children with Charcot-Marie-Tooth Disease.: This study highlights the use of 3D printing with Nylon 12 for creating custom ankle-foot orthoses, enhancing comfort and functionality for pediatric patients. (Wojciechowski et al., 2022).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Habiba Bougherara et al.
Journal of biomedical materials research. Part A, 82(1), 27-40 (2007-02-01)
A new biomimetic composite hip prosthesis (stem) was designed to obtain properties similar to those of the contiguous bone, in particular stiffness, to allow normal loading of the surrounding femoral bone. This normal loading would reduce excessive stress shielding, known
Kosuke Tomita et al.
Biotechnology letters, 25(20), 1743-1746 (2003-11-25)
A thermophilic bacterium, identified as a neighboring species to Geobacillus thermocatenulatus, having a growth optimum at 55 degrees C and, capable of degrading nylon 12, was isolated from soil by enrichment culture technique at 60 degrees C. At this temperature
K R Williams et al.
Biomaterials, 8(4), 254-258 (1987-07-01)
Implantation of three different polymeric composite materials in the rat middle ear showed that the nylon prosthesis was encapsulated more rapidly than the two poly(methyl methacrylate) based implants. Several PMMA based implants were only partially covered at the 6 mnth
N H Stoffers et al.
Food additives and contaminants, 20(4), 410-416 (2003-05-31)
Analytical methods for the determination of laurolactam--the monomer of nylon 12--as well as the cyclic dimer and trimer were established. High performance liquid chromatography using ultraviolet (HPLC-UV) and mass spectrometric detection (HPLC-MS) were both found suitable to identify and quantify
N H Stoffers et al.
Food additives and contaminants, 20(10), 949-959 (2003-11-05)
The migration of laurolactam and cyclic di- and trimer of nylon 12 was assessed using three different films and five food simulants (olive oil, isooctane, 95% ethanol, 50% ethanol, water). Substitute test conditions for migration into olive oil according to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service