Skip to Content
Merck
All Photos(1)

Documents

180548

Sigma-Aldrich

2,4-Heptadienal

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH=CHCH=CHCHO
CAS Number:
Molecular Weight:
110.15
Beilstein:
1699244
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

liquid

refractive index

n20/D 1.534 (lit.)

bp

84-84.5 °C (lit.)

density

0.881 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC\C=C\C=C\C=O

InChI

1S/C7H10O/c1-2-3-4-5-6-7-8/h3-7H,2H2,1H3/b4-3+,6-5+

InChI key

SATICYYAWWYRAM-VNKDHWASSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Heptadienal is a specific marker of oxidative rancidity. It is also a potential algal metabolite.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yunyun Zhao et al.
Journal of environmental sciences (China), 25(12), 2361-2366 (2014-03-22)
A significant outbreak of fishy odor occurred in a reservoir located in Inner Mongolia, China, in the winter of 2011, and the odor rating, algal density and concentrations of some potential odorous compounds were monitored over a period of two
Satomi Shimoyoshi et al.
Nutrients, 11(7) (2019-07-25)
Sesame lignans, which are biologically active compounds present in sesame seeds and oil, are known to have neuroprotective effects in several models of brain dysfunction. However, the effects of sesame lignans on age-related brain dysfunction are not clear and were
Ai Suzuki-Iwashima et al.
Food chemistry, 334, 127588-127588 (2020-07-30)
A mixture of sphingoid bases (SPGs) was prepared from butter serum, a by-product of anhydrous milk fat production. The mixture comprised seven types of SPGs with C16 to C19 alkyl chains. These milk SPGs inhibited the oxidation of fish oil
Charles Vidoudez et al.
Marine drugs, 9(4), 500-513 (2011-07-07)
Polyunsaturated aldehydes (PUA) are supposed to play critical roles in chemically-mediated plankton interactions. Laboratory studies suggest that they act as mediators of chemical defense and chemical communication. PUA are oxylipins containing an α,β,γ,δ-unsaturated aldehyde structure element and are mainly found
Wakana Onuma et al.
Nutrition and cancer, 69(6), 855-861 (2017-07-19)
Colorectal cancer is one of the leading causes of death worldwide. Reactive oxygen species produce oxidative stress and contribute to colorectal carcinogenesis. Because dietary citrus has been shown to reduce oxidative stress, we investigated the effects of citrus peel extract

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service