Skip to Content
Merck
All Photos(2)

Key Documents

178497

Sigma-Aldrich

L-Selectride® solution

1.0 M in THF

Synonym(s):

Lithium tri-sec-butylborohydride solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li[CH(CH3)CH2CH3]3BH
CAS Number:
Molecular Weight:
190.10
Beilstein:
4007327
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reagent type: reductant

concentration

1.0 M in THF

density

0.89 g/mL at 25 °C

SMILES string

[Li+].[H][B-](C(C)CC)(C(C)CC)C(C)CC

InChI

1S/C12H28B.Li/c1-7-10(4)13(11(5)8-2)12(6)9-3;/h10-13H,7-9H2,1-6H3;/q-1;+1

InChI key

ACJKNTZKEFMEAK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Selectride® is an air and water sensitive solution mainly used as a bulky stereoselective hydride reducing agent.

Application

L-Selectride® can be used as a reagent for:
  • The stereoselective reduction of ketone to alcohol.
  • The selective demethylation of tert alcohol.

L-Selectride® solution may be used in the O-demethylation and N-demethylation of opium alkaloids and their derivatives.
  • Reducing agent

Reagent for:
  • Enantioselective synthesis of a-amino acids via reduction of N-tert-butanesulfinyl ketimine esters
  • Diastereoselective reduction reactions
  • Hydride reduction of the Danishefsky pyranones
  • Asymmetric reductive aldol reaction of enones with a-alkoxy aldehydes
  • Synthesis of alkanols by carbonylation reactions

Legal Information

Selectride is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Carc. 2 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective synthesis of a novel pseudopeptide hapten for the generation of hydrolytic catalytic antibodies
Rodriguez AC, et al.
Tetrahedron Asymmetry, 15(12), 1847-1855 (2004)
L-Selectride as a General Reagent for the O-Demethylation and N-Decarbomethoxylation of Opium Alkaloids and Derivatives1.
Coop A, et al.
The Journal of Organic Chemistry, 63(13), 4392-4396 (1998)
Li JJ, et al.
Modern Organic Synthesis in the Laboratory (2007)
Lithium Tri-sec-butylborohydride
Hubbard JL and Dake G.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2012)
A General and Simple Diastereoselective Reduction by L-Selectride: Efficient Synthesis of Protected (4S, 5S)-Dihydroxy Amides.
Yin B, et al.
Molecules (Basel), 15(4), 2771-2781 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service