Skip to Content
Merck
All Photos(1)

Key Documents

175927

Sigma-Aldrich

p-Xylene-d10

99 atom % D

Synonym(s):

1,4-Dimethylbenzene-d10

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D4(CD3)2
CAS Number:
Molecular Weight:
116.23
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99 atom % D

Assay

≥98.5%

form

liquid

expl. lim.

0.34-6.3 % (lit.)

technique(s)

GC/MS: suitable
NMR: suitable

refractive index

n20/D 1.492 (lit.)

bp

135 °C (lit.)

mp

13 °C (lit.)

density

0.948 g/mL at 25 °C (lit.)

mass shift

M+10

SMILES string

[2H]c1c([2H])c(c([2H])c([2H])c1C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C8H10/c1-7-3-5-8(2)6-4-7/h3-6H,1-2H3/i1D3,2D3,3D,4D,5D,6D

InChI key

URLKBWYHVLBVBO-ZGYYUIRESA-N

General description

p-Xylene-d10 is a deuterated derivative of p-xylene. It has an isotopic purity of 98atom%D. A series of solutions of p-xylene dissolved in the first 12 members of the 4-n-alkyloxy-4′-cyanobiphenyls (NOCB) were prepared. Principal components Szz and Sxx - Syy of the second rank orientational ordering matrix of these solutions were determined by deuterium NMR analysis. It is an anthropogenic volatile organic compounds (VOC), which is useful for the formation of secondary organic aerosol (SOA).

Application

p-Xylene-d10 may be used as an internal standard (IS) for GC/MS analysis of volatile organic compounds in the emissions from a landfill.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evolution of the complex refractive index in the near UV spectral region in ageing secondary organic aerosol.
Flores JM, et al.
Atmospheric Chemistry and Physics Discussions, 14, 4149-4187 (2014)
The crystal structures of m-xylene and p-xylene, C8D10, at 4.5 K.
Ibberson RM, et al.
Journal of Molecular Structure, 524(1), 121-128 (2000)
Characterisation of odorants emissions from landfills by SPME and GC/MS.
Davoli E, et al.
Chemosphere, 51(5), 357-368 (2003)
Dependence of solute orientational order on the structure and orientational order of the liquid crystal solvent. A deuterium NMR study of p-xylene-d10 dissolved in a series of 4-n-alkyloxy-4'-cyanobiphenyls.
Emsley JW, et al.
Liq. Cryst., 5(3), 953-967 (1989)
Zhihui Feng et al.
Food chemistry, 285, 347-354 (2019-02-25)
Tea aroma is determined by the nature of the plant, the production processes, and many other factors influencing its formation and release. The objective of this study was to investigate the impact of manufacturing processes on the aroma composition of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service