Skip to Content
Merck
All Photos(1)

Key Documents

175862

Sigma-Aldrich

Acetone-d6

"100%", 99.95 atom % D

Synonym(s):

Hexadeuteroacetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3COCD3
CAS Number:
Molecular Weight:
64.12
Beilstein:
1702935
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

vapor density

2 (vs air)

Quality Level

vapor pressure

14.39 psi ( 55 °C)
184 mmHg ( 20 °C)
3.67 psi ( 20 °C)

isotopic purity

99.95 atom % D

Assay

≥99% (CP)

form

liquid

expl. lim.

13.2 %

packaging

ampule of 0.5, 0.75 mL, 5 g
glass bottle of 25 g

technique(s)

NMR: suitable

impurities

≤0.0500% water
water

refractive index

n20/D 1.355 (lit.)

bp

55.5 °C (lit.)

mp

−93.8 °C (lit.)

density

0.872 g/mL at 25 °C (lit.)

mass shift

M+6

SMILES string

[2H]C([2H])([2H])C(=O)C([2H])([2H])[2H]

InChI

1S/C3H6O/c1-3(2)4/h1-2H3/i1D3,2D3

InChI key

CSCPPACGZOOCGX-WFGJKAKNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetone-d6 (CD3COCD3, Hexadeuteroacetone) is the hexadeuterated form of acetone. It is 100% NMR (Nuclear Magnetic Resonance) solvent. It is widely employed in high resolution NMR studies due to its high chemical and isotopic purity. Absorption spectra of CD3COCD3 samples cooled in molecular jet (jet-cooled) have been evaluated in the range of 51000-54000cm-1.[1] Collision of its molecular ions with a perdeuterated surface (C20D41 SAu) was investigated by ion-surface collision spectroscopy.[2] The Infrared and Raman spectra of acetone-d6 have been analyzed.4 It undergoes H-2H exchange reaction with acetone d0 to afford acetone d1 and d5.[3]
Acetone-d6 can be prepared by reacting heavy water with propyne-d4 in the presence of mercury catalyst.[4]

Application

Acetone-d6 has been used as a solvent for the NMR (Nuclear Magnetic Resonance) spectral investigation of (S)-2-(6-((3-((15-carboxypentadecyl)disulfanyl)propoxy)carbonyloxy)benzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid.[5]

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldol condensation of acetone/acetone d6 over magnesium oxide and lanthanum oxide.
Zhang G, et al.
Applied Catalysis, 40, 183-190 (1988)
Reactive ion-surface collisions: Application of ionized acetone-d6, DMSO-d6 and pyridine-d5 as probes for the characterization of self-assembled monolayer films on gold.
Kane TE, et al.
Org. Mass Spectrom., 28(12), 1665-1673 (1993)
The (n0-3s) Rydberg state of acetone: absorption spectroscopy of jet-cooled acetone and acetone-d6.
Gaines GA, et al.
The Journal of Physical Chemistry, 92(10), 2762-2766 (1988)
Elena Dubikovskaya et al.
Methods in enzymology, 538, 107-134 (2014-02-18)
The ability of white and brown adipose tissue to efficiently take up long-chain fatty acids is key to their physiological functions in energy storage and thermogenesis, respectively. Several approaches have been taken to determine uptake rates by cultured cells and
Neue Synthese von Aceton-d6.
Hunziker H, et al.
Helvetica Chimica Acta, 45(1), 59-62 (1962)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service