Skip to Content
Merck
All Photos(1)

Key Documents

175706

Sigma-Aldrich

Aniline-d7

98 atom % D

Synonym(s):

Heptadeuteroaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D5ND2
CAS Number:
Molecular Weight:
100.17
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

refractive index

n20/D 1.5824 (lit.)

bp

184 °C (lit.)

mp

-6 °C (lit.)

density

1.098 g/mL at 25 °C

mass shift

M+7

storage temp.

2-8°C

SMILES string

[2H]N([2H])c1c([2H])c([2H])c([2H])c([2H])c1[2H]

InChI

1S/C6H7N/c7-6-4-2-1-3-5-6/h1-5H,7H2/i1D,2D,3D,4D,5D/hD2

InChI key

PAYRUJLWNCNPSJ-LQHBDRBESA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thorben Vockenberg et al.
Environmental science. Processes & impacts, 22(8), 1678-1687 (2020-07-09)
Microplastics are ubiquitous in the environment. Due to still rising global production, the emission of polymers into the environment and the abundance of microplastics have increased accordingly. Due to the long mineralization processes of microplastics, distribution in all compartments can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service