Skip to Content
Merck
All Photos(3)

Documents

175404

Sigma-Aldrich

4-Isopropylphenol

98%

Synonym(s):

p-Cumenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHC6H4OH
CAS Number:
Molecular Weight:
136.19
Beilstein:
1363564
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Assay

98%

form

crystals

bp

212-213 °C (lit.)

mp

59-61 °C (lit.)

SMILES string

CC(C)c1ccc(O)cc1

InChI

1S/C9H12O/c1-7(2)8-3-5-9(10)6-4-8/h3-7,10H,1-2H3

InChI key

YQUQWHNMBPIWGK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sonochemical degradation of 4-Isopropylphenol has been studied.

Application

4-Isopropylphenol was used in the synthesis of vinyl compounds.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - Pensky-Martens closed cup

Flash Point(C)

110 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahdi Chiha et al.
Ultrasonics sonochemistry, 17(5), 773-782 (2010-04-15)
Sonochemical degradation of phenol (Ph), 4-isopropylphenol (4-IPP) and Rhodamine B (RhB) in aqueous solutions was investigated for a large range of initial concentrations in order to analyze the reaction kinetics. The initial rates of substrate degradation and H(2)O(2) formation as
Frédéric L P Gabriel et al.
Chemistry & biodiversity, 4(9), 2123-2137 (2007-09-25)
Sphingobium xenophagum Bayram is capable of metabolizing 4-alkoxyphenols and endocrine disruptive alpha-quaternary 4-nonylphenols by an ipso-substitution mechanism that involves ring hydroxylation at the site of the substituent. Here, we show that Bayram's ipso-hydroxylating activity was able to transform also bisphenol
David J Hopper et al.
Applied and environmental microbiology, 69(6), 3650-3652 (2003-06-06)
4-ethylphenol methylenehydroxylase from Pseudomonas putida JD1 acts by dehydrogenation of its substrate to give a quinone methide, which is then hydrated to an alcohol. It was shown to be active with a range of 4-alkylphenols as substrates. 4-n-propylphenol, 4-n-butylphenol, chavicol
Ye Eun Park et al.
Nutrients, 12(1) (2020-01-23)
Probiotics can improve the intestinal environment by enhancing beneficial bacteria to potentially regulate lipid levels; however, the underlying mechanisms remain unclear. The aim of this study was to investigate the effect of Lactobacillus plantarum Q180 (LPQ180) on postprandial lipid metabolism
D C Thompson et al.
Chemical research in toxicology, 8(1), 55-60 (1995-01-01)
The oxidative metabolism and toxicity of the para isomers of methylphenol (cresol), ethylphenol, and isopropylphenol were studied using male Sprague-Dawley rat liver microsomes and precision-cut liver slices. Reactive intermediates from each compound were trapped using radiolabeled glutathione and were detected

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service