Skip to Content
Merck
All Photos(1)

Key Documents

17343

Sigma-Aldrich

Bromhexine hydrochloride

≥98.0% (AT)

Synonym(s):

2-Amino-3,5-dibromo-N-cyclohexyl-N-methylbenzylamine hydrochloride, N-(2-Amino-3,5-dibromobenzyl)-N-methylcyclohexylamine hydrochloride, Bromohexine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20Br2N2 · HCl
CAS Number:
Molecular Weight:
412.59
Beilstein:
4848376
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (AT)

form

powder

functional group

amine
bromo

SMILES string

Cl.CN(Cc1cc(Br)cc(Br)c1N)C2CCCCC2

InChI

1S/C14H20Br2N2.ClH/c1-18(12-5-3-2-4-6-12)9-10-7-11(15)8-13(16)14(10)17;/h7-8,12H,2-6,9,17H2,1H3;1H

InChI key

UCDKONUHZNTQPY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bromhexine hydrochloride is a bronchial mucolytic.

Application

Bromhexine hydrochloride was used in the secretion of pancreatic juice of low viscosity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatsuhiro Tsujimoto et al.
Alcoholism, clinical and experimental research, 29(12 Suppl), 272S-276S (2005-12-31)
Abstinence is a prerequisite for the treatment of alcoholic chronic pancreatitis, but there are patients who have repeated attacks because of their inability to abstain and the consequent congestive effects of the continued alcohol intake on pancreatic juice. Bromhexine hydrochloride
Mladen Stojanov et al.
International journal of pharmaceutics, 422(1-2), 349-355 (2011-10-27)
Bromhexine hydrochloride (bromhexine) is a mucolytic agent with very low aqueous solubility. However, with addition of cyclodextrins (CD) to the formulation, this disadvantage may be limited and therapeutic doses of bromhexine in solution can be achieved. The interaction of bromhexine
Marcello Nicoletti et al.
Natural product research, 24(14), 1325-1329 (2010-08-31)
The utilisation of NMR fingerprinting is proposed as a rapid, available and reliable method to determine the contamination of herbal products. The presence of nimesulide has been reported recently as the contaminant of P.C. 28 Plus, a product based on
Superior mesenteric artery aneurysm: importance of sonography as the primary imaging technique for detection.
Radu Badea et al.
Journal of ultrasound in medicine : official journal of the American Institute of Ultrasound in Medicine, 29(10), 1503-1506 (2010-09-30)
Matthew E Spector et al.
Head & neck, 34(12), 1727-1733 (2012-01-17)
Despite better prognosis, there is a group of oropharyngeal squamous cell carcinoma (SCC) human papillomavirus (HPV)+ patients who experience treatment failure and succumb to distant metastasis. Seventy-eight previously untreated patients nested in a concurrent chemoradiation protocol were reviewed to correlate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service