Skip to Content
Merck
All Photos(1)

Documents

166804

Sigma-Aldrich

Dibenzyl azodicarboxylate

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OCON=NCOOCH2C6H5
CAS Number:
Molecular Weight:
298.29
Beilstein:
2298734
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

90%

form

solid

mp

43-47 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C(OCc1ccccc1)\N=N\C(=O)OCc2ccccc2

InChI

1S/C16H14N2O4/c19-15(21-11-13-7-3-1-4-8-13)17-18-16(20)22-12-14-9-5-2-6-10-14/h1-10H,11-12H2/b18-17+

InChI key

IRJKSAIGIYODAN-ISLYRVAYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibenzyl azodicarboxylate undergoes[4+2] cycloaddition reaction with glycal to yield 2-aminoglycosides.

Application

Dibenzyl azodicarboxylate was used as electrophilic reagent in the synthesis of C-glycosyl α-amino acids via proline-catalyzed α-amination of C-glycosylalkyl aldehydes. It was used in enantioselective synthesis of optically active pyrazolidine derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shengming Ma et al.
Organic letters, 6(13), 2193-2196 (2004-06-18)
[reaction: see text] Optically active pyrazolidine derivatives have been constructed by the Cu- and Pd-catalyzed asymmetric one-pot tandem addition-cyclization reaction of 2-(2',3'-dienyl)-beta-ketoesters, organic halides, and dibenzyl azodicarboxylate. The absolute configurations of the final products depend on the structure of the
Andrea Nuzzi et al.
Organic letters, 10(20), 4485-4488 (2008-09-16)
Non-natural axially and equatorially linked C-glycosyl alpha-amino acids (glycines, alanines, and CH2-serine isosteres) with either S or R alpha-configuration were prepared by D- and L-proline-catalyzed (de >95%) alpha-amination of C-glycosylalkyl aldehydes using dibenzyl azodicarboxylate as the electrophilic reagent.
[4+ 2] Cycloaddition reaction of dibenzyl azodicarboxylate and glycals.
Leblanc Y, et al.
Journal of the American Chemical Society, 111(8), 2995-3000 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service