Skip to Content
Merck
All Photos(1)

Key Documents

164542

Sigma-Aldrich

Ethyl 3-furoate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O3
CAS Number:
Molecular Weight:
140.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
98%

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.46 (lit.)

bp

93-95 °C/35 mmHg (lit.)

density

1.038 g/mL at 25 °C (lit.)

functional group

ester

storage temp.

2-8°C

SMILES string

CCOC(=O)c1ccoc1

InChI

1S/C7H8O3/c1-2-10-7(8)6-3-4-9-5-6/h3-5H,2H2,1H3

InChI key

LOFDXZJSDVCYAS-UHFFFAOYSA-N

General description

Ethyl 3-furoate undergoes regioselective palladium(0)-catalyzed arylation reaction with aryl bromides.

Application

Ethyl 3-furoate was used as starting reagent for the synthesis of ethyl 2,3-bis(trifluoromethyl)-7-oxabicyclo[2,2,1]hepta-2,5-diene-5-carboxylate and 4-(1-hydroxy-1-methyl-ethyl)-furan-2-sulfonamide.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel Synthesis of 1-(1, 2, 3, 5, 6, 7-Hexahydro-s-indacen-4-yl)-3-[4-(1-hydroxy-1-methyl-ethyl)-furan-2-sulfonyl] urea, an Anti-inflammatory Agent.
Urban FJ, et al.
Synthetic Communications, 33(12), 2029-2043 (2003)
Some reactions of 3, 4-bis (trifluoromethyl) furan and its precursor, 2, 3-bis (trifluoromethyl)-7-oxabicyclo [2, 2, 1] hepta-2, 5-diene: novel isocoumarin formation from thermal reaction of the furan with ethyl propynoate.
Abubakar AB, et al.
Journal of Fluorine Chemistry, 56(3), 359-371 (1992)
Bobby Glover et al.
Organic letters, 5(3), 301-304 (2003-01-31)
[reaction: see text] The regioselective palladium(0)-catalyzed arylation of 3-furoate and 3-thiophenecarboxylate esters with aryl bromides is described. Conditions were developed that allow for the selective synthesis of either 2-aryl or 5-aryl products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service