All Photos(1)
About This Item
Linear Formula:
FC6H4OCH3
CAS Number:
Molecular Weight:
126.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
form
liquid
refractive index
n20/D 1.488 (lit.)
bp
158 °C/743 mmHg (lit.)
density
1.104 g/mL at 25 °C (lit.)
functional group
fluoro
SMILES string
COc1cccc(F)c1
InChI
1S/C7H7FO/c1-9-7-4-2-3-6(8)5-7/h2-5H,1H3
InChI key
MFJNOXOAIFNSBX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
3-Fluoroanisole was used in the synthesis of 4-fluoro-5,6-dihydroxytryptamine. It was also used in the synthesis of 3-fluoro- and 5-fluoronoradrenaline.
Signal Word
Warning
Hazard Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
111.2 °F - closed cup
Flash Point(C)
44 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis and EPR investigations of fluorocatecholamines.
Stegmann HB, et al.
Journal of the Chemical Society. Perkin Transactions 1, 2(3), 547-555 (1994)
M Kawase et al.
Journal of medicinal chemistry, 33(8), 2204-2211 (1990-08-01)
The 5,6-dihydroxytryptamine (5,6-DHT) derivatives 4-fluoro- and 7-fluoro-5,6-DHTs (26a,b) and 4,7-difluoro-5,6-DHT (26c) were synthesized from 3-fluoroanisole (1) and 1,4-difluoro-2,3-dimethoxybenzene (13), respectively. Efficient methods were developed for the conversion of 1 to 4-fluoro- and 7-fluoro-5,6-bis(benzyloxy)indoles (12a,b, respectively), and 13 to 4,7-difluoro-5,6-[( diphenylmethylene)dioxy]indole
Oriol Planas et al.
Science (New York, N.Y.), 367(6475), 313-317 (2020-01-18)
Bismuth catalysis has traditionally relied on the Lewis acidic properties of the element in a fixed oxidation state. In this paper, we report a series of bismuth complexes that can undergo oxidative addition, reductive elimination, and transmetallation in a manner
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service