Skip to Content
Merck
All Photos(1)

Key Documents

160385

Sigma-Aldrich

Phthalimide potassium salt

98%

Synonym(s):

1,3-Dihydro-1,3-dioxoisoindole potassium salt, Potassium phthalimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4KNO2
CAS Number:
Molecular Weight:
185.22
Beilstein:
3598719
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

>300 °C (lit.)

solubility

water: soluble 50 mg/mL, clear to slightly hazy, colorless to yellow

functional group

imide

SMILES string

[K]N1C(=O)c2ccccc2C1=O

InChI

1S/C8H5NO2.K/c10-7-5-3-1-2-4-6(5)8(11)9-7;/h1-4H,(H,9,10,11);/q;+1/p-1

InChI key

FYRHIOVKTDQVFC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Condensation of phthalimide potassium with organic halide in dimethylformamide has been reported. Reaction of potassium phthalimide and sulfur monochloride in petroleum ether has been studied.
Phthalimide potassium salt was employed as organocatalyst for the cyanosilylation of various carbonyl compounds under extremely mild conditions. It was also employed as reagent for the transformation of allyl- and alkyl halides into protected primary amines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

346.3 °F - closed cup

Flash Point(C)

174.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 48, 3043-3043 (2007)
Reactions of Phthalimide and Potassium Phthalimide with Sulfur Monochloride.
Kalnins MV.
Canadian Journal of Chemistry, 44(17), 2111-2113 (1966)
An improved procedure for the condensation of potassium phthalimide with organic halides.
Sheehan JC and Bolhofer WA.
Journal of the American Chemical Society, 72(6), 2786-2788 (1950)
Activation of trimethylsilyl cyanide by potassium phthalimide for facile synthesis of TMS-protected cyanohydrins.
Dekamin MG and Karimi Z.
Journal of Organometallic Chemistry, 694(12), 1789-1794 (2009)
Palladium-catalyzed direct functionalization of imidazolinone: synthesis of dibromophakellstatin.
Jianming Lu et al.
Journal of the American Chemical Society, 129(25), 7768-7769 (2007-06-02)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service