Skip to Content
Merck
All Photos(3)

Key Documents

156515

Sigma-Aldrich

2,4,5-Trichlorophenol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3C6H2OH
CAS Number:
Molecular Weight:
197.45
Beilstein:
607569
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

bp

248 °C/740 mmHg (lit.)

mp

67-69 °C (lit.)

functional group

chloro

SMILES string

Oc1cc(Cl)c(Cl)cc1Cl

InChI

1S/C6H3Cl3O/c7-3-1-5(9)6(10)2-4(3)8/h1-2,10H

InChI key

LHJGJYXLEPZJPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2,4,5-Trichlorophenol (2,4,5-TCP) is employed as a raw material in the preparation of various biocides.[1]

Application

2,4,5-trichlorophenol can be used as a starting material to synthesize:
  • Hexachlorophene, a fungicide, by the reaction with formaldehyde in the presence of concentrated H2SO4.[1][2]
  • 2,4,5-trichlorophenoxyacetic acid (2,4,5-T), a herbicide.[1][3]
  • N-Benzyl-2-(2,4,5-trichlorophenoxy)acetamide, which is used as an intermediate to prepare 4-benzyl-6,7-dichloro-2H-benzo[b][1,4]oxazin-3(4H)-one via Smiles rearrangement.[4]

The product has been used to study its sorption onto montmorillonite based sorbents.

Disclaimer

“The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others."

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

271.4 °F - closed cup

Flash Point(C)

133.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Organic Chemistry: With Biological Applications
Organic Chemistry (2011)
Microwave-assisted Synthesis of 2H-Benzo [b][1, 4] oxazin-3 (4H)-ones and 1H-Pyrido [2, 3-b][1, 4] oxazin-2 (3H)-ones via Smiles Rearrangement
Hua Zuo, et al.
Bulletin of the Korean Chemical Society,, 29(7), 1379-1385 (2008)
Photo-assisted degradation of 2, 4, 5-trichlorophenol by Electro-Fe (II)/Oxone{\textregistered} process using a sacrificial iron anode: Performance optimization and reaction mechanism
Wang YR and Chu Wei
Chemical Engineering Journal, 215, 643-650 (2013)
The Lethal Interaction and Formation of a Lipophilic Ternary Complex between 2, 4, 5-Trichlorophenol and the Cu (II)- Bis (1, 10-phenanthroline) Complex
Zhu Ben-Zhan
Chemical Research in Toxicology, 14(2), 222-227 (2001)
Qiao Sun et al.
Environmental science & technology, 41(16), 5708-5715 (2007-09-19)
Density functional theory (DFT) calculations have been carried out to explore the potential energy surface (PES) associated with the gas-phase reaction between 2,4,5-trichlorophenol and CuO. A gas-phase model was constructed to account, from a theoretical perspective, for the most important

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service